Skip to Content
Merck
All Photos(1)

Documents

MAB3564

Sigma-Aldrich

Anti-Sodium/Iodine Symporter Antibody, a.a. 625-643, clone FP5A

clone FP5A, Chemicon®, from mouse

Synonym(s):

Na+/I- Symporter, NIS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

FP5A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC5A5(6528)

Specificity

Sodium/Iodide Symporter (NIS). The epitope is thought to be in the region of amino acids 625-643 of human NIS. The antibody may show some reactivity to Maltose Binding Protein.

Immunogen

Epitope: a.a. 625-643
Maltose Binding Protein fusion protein corresponding to amino acids 468-643 of human NIS (C-terminus).

Application

Detect Sodium/Iodine Symporter using this Anti-Sodium/Iodine Symporter Antibody, a.a. 625-643, clone FP5A validated for use in IH, IH(P) & WB.
Immunoblotting. The antibody recognizes a major band at ~97 kDa and minor bands at 15 kDa, 30 kDa, 68 kDa and 160 kDa.

Immunohistochemistry. The antibody works on paraffin embedded tissue sections which have been subjected to antigen retrieval with 10 mM citrate buffer steam for 30 minutes.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annie Yang et al.
Journal for immunotherapy of cancer, 11(4) (2023-04-06)
Gastric cancer (GC) that metastasizes to the peritoneum is fatal. CF33 and its genetically modified derivatives show cancer selectivity and oncolytic potency against various solid tumors. CF33-hNIS and CF33-hNIS-antiPDL1 have entered phase I trials for intratumoral and intravenous treatments of
Christina Schug et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(19), 5997-6008 (2019-06-15)
The innate tumor homing potential of mesenchymal stem cells (MSCs) has been used for a targeted delivery of the theranostic sodium iodide symporter (NIS) transgene into solid tumors. We have previously shown that external beam radiotherapy (EBRT) results in the
Carolin Kitzberger et al.
Molecular therapy oncolytics, 30, 238-253 (2023-09-13)
New treatment strategies are urgently needed for glioblastoma (GBM)-a tumor resistant to standard-of-care treatment with a high risk of recurrence and extremely poor prognosis. Based on their intrinsic tumor tropism, adoptively applied mesenchymal stem cells (MSCs) can be harnessed to
Danielle L Gainor et al.
JAMA otolaryngology-- head & neck surgery, 141(8), 739-744 (2015-06-05)
Sodium iodide symporter (NIS) expression in adenoid cystic carcinoma (ACC) has not been fully elucidated in the literature, and it is unclear whether radioactive iodine may be a potential therapeutic modality. To our knowledge, the present study includes the largest
Rebekka Spellerberg et al.
Molecular therapy oncolytics, 27, 272-287 (2022-12-03)
Sodium iodide symporter (NIS) gene transfer for active accumulation of iodide in tumor cells is a powerful theranostic strategy facilitating both diagnostic and therapeutic application of radioiodide. In glioblastoma (GBM), the blood-brain barrier (BBB) presents an additional delivery barrier for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service