Skip to Content
Merck
All Photos(1)

Key Documents

MAB3562

Sigma-Aldrich

Anti-Sodium/Iodine Symporter Antibody, clone 2-2

clone 2-2, Chemicon®, from mouse

Synonym(s):

Na+/I- Symporter, NIS

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$743.00

$743.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2-2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC5A5(6528)

Specificity

Sodium/Iodide Symporter (NIS)

Immunogen

Synthetic peptide corresponding to amino acids 37-54 of human NIS.

Application

Anti-Sodium/Iodine Symporter Antibody, clone 2-2 detects level of Sodium/Iodine Symporter & has been published & validated for use in IH & WB.
Immunoblotting. The antibody recognizes a major band at ~97 kDa and minor bands at 15 kDa, 30 kDa, 68 kDa and 160 kDa.

Immunohistochemistry on frozen tissues.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Micali et al.
The Journal of endocrinology, 216(2), 125-133 (2012-11-03)
Testicular cancer is the most frequent cancer in young men. The large majority of patients have a good prognosis, but in a small group of tumors, the current treatments are not effective. Radioiodine is routinely used in the treatment of
Elisa Sala et al.
Molecular cancer research : MCR, 6(5), 751-759 (2008-05-07)
BRAF-activating mutations have been reported in several types of cancer, including melanoma ( approximately 70% of cases), thyroid (30-70%), ovarian (15-30%), and colorectal cancer (5-20%). Mutant BRAF has constitutive kinase activity and causes hyperactivation of the mitogen-activated protein kinase pathway.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service