Skip to Content
Merck
All Photos(4)

Key Documents

MAB353B

Sigma-Aldrich

Anti-Nestin Antibody, clone rat-401, Biotin Conjugate

clone rat-401, from mouse, biotin conjugate

Synonym(s):

rat401, NES, Nbla00170, FLJ21841

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$671.00

$671.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
$671.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$671.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

rat-401, monoclonal

species reactivity

mouse, rat

should not react with

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NES(10763)
mouse ... Nes(18008)
rat ... Nes(25491)

General description

Nestin, a large intermediate filament protein (class Type VI) expressed during development and in myotendinous and neuromuscular junctions. Nestin expression is restricted, typically disappearing by E18. Nestin is thought to be a reasonable neuronal marker; however, recent studies have found nestin expression in other cell types such as endothelial cells (Mokry and Nemecek, 1998). Nestin identifies the most primitive neuroepithelium but also identifies many other embryonic tissues, so it is not specific for CNS. Nestin expression is seen in almost all GBMs (Glioblastoma mulitformes) and many melanomas (both primary and metastatic) but not in any metastatic carcinomas.

Specificity

This antibody recognizes both rat and mouse Nestin. Antibody is not reactive with human Nestin protein.

Immunogen

Embryonic rat spinal cord

Application

Detect Nestin using this mouse monoclonal antibody, Anti-Nestin Antibody, clone rat-401, Biotin Conjugate validated for use in ICC & IHC.
Evaluated by Immunocytochemistry in rat hippocampal neural stem cells (SCR022).
Immunocytochemsitry Analysis: A 1:200 dilution of this antibody detected Nestin in rat hippocampal neural stem cells (SCR022).
Evaluated by Immunocytochemistry in ReNcell VM human neural progenitor cells (SCC008).
Immunocytochemsitry Analysis: A 1:200 dilution of this antibody did not detect Nestin in ReNcell VM human neural progenitor cells (SCC008). Evaluated by Immunohistochemistry in rat hindbrain brain tissue.
Immunohistochemsitry Analysis: A 1:50 dilution of this antibody detected Nestin in rat hindbrain brain tissue.
Research Category
Stem Cell Research

Neuroscience
Research Sub Category
Developmental Neuroscience

Neural Stem Cells

Quality

Evaluated by Immunocytochemistry in mouse cortical neural stem cells (SCR029). Immunocytochemsitry Analysis: A 1:200 dilution of this antibody detected Nestin in mouse cortical neural stem cells (SCR029).

Target description

200-220 kDa calculated

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 conjugated to Biotin in PBS with 0.1% sodium azide and 15mg/ml BSA.

Storage and Stability

Maintain refrigerated at 2-8ºC protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Mouse cortical neural stem cells (SCR029)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dandan Gu et al.
Molecular medicine reports, 16(6), 7993-8002 (2017-10-07)
Specification of distinct cell types from Müller glial cells is key to the potential application of endogenous repair in retinal regeneration. Sonic hedgehog (SHH) has been established as a potent mitogen for rat Müller glial cells, which also induces Müller

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service