Skip to Content
Merck
All Photos(1)

Key Documents

MAB348-AF555

Sigma-Aldrich

Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, Alexa Fluor 555

clone 22C11, from mouse, ALEXA FLUOR 555

Synonym(s):

Amyloid beta A4 protein, ABPP, Alzheimer disease amyloid protein, Amyloid precursor protein, APP, APPI, Cerebral vascular amyloid peptide, CVAP, PN-II, PreA4, Protease nexin-II, PN-II, N-APP, Soluble APP-alpha, S-APP-alpha, Soluble APP-beta, S-APP-beta,

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$888.00

$888.00


Estimated to ship on16 April 2025



Select a Size

Change View
100 μL
$888.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$888.00


Estimated to ship on16 April 2025


biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22C11, monoclonal

species reactivity

canine, human, mouse, rat, porcine, monkey, fish

technique(s)

immunocytochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

General description

Amyloid beta A4 protein (UniProt P05067; also known as ABPP, Alzheimer disease amyloid protein, Amyloid precursor protein, APP, APPI, Cerebral vascular amyloid peptide, CVAP, PN-II, PreA4, Protease nexin-II) is encoded by the APP (also known as A4, AD1) gene (Gene ID 351) in human. Amyloid precursor protein (APP) is initially produced with a signal peptide sequence (a.a. 1-17), the removal of which yields the mature protein with a large extracellular portion (a.a. 18-699), followed by a transmembrane segment (a.a. 700-723) and a cytoplasmic (a.a. 724-770) tail. APP can be further processed by the α-, β-, and γ-secretases in two alternative processing pathways. In the non-amyloidogenic pathway, APP is first cleaved by the plasma membrane-localized α-secretase to generate an N-terminal extracellular sAPPα fragment (a.a. 18-687) and a membrane-bound C-terminal fragment C83 (CTFα), which can be further cleaved by γ-secretase to produce a non-toxic small peptide p3 and a cytoplasmic APP intracellular domain (AICD). In the amyloidogenic pathway, APP undergoes β-cleavage in BACE-1 (β-site APP-cleaving enzyme)-enriched endosomes to generate an N-terminal extracellular sAPPβ fragment (a.a. 18-671) and a membrane-bound C-terminal fragment C99 (CTFβ). Subsequent cleavage of C99 by γ-secretase releases the amyloid β peptide (Aβ) and AICD. Aβ accumulation in the cortical and hippocampal regions of the brain is a major pathological feature of Alzheimer′s disease (AD).

Specificity

Clone 22C11 reacts with pre-A4. The antibody recognizes amino acids 66-81 of the N-terminus on the pre-A4 molecule (Hilbich, C., et al. (1993). J. Biol. Chem. 268(35):26571-26577). Clone 22C11 recognizes all three isoforms of APP, immature ~110kDa, sAPP ~120kDa, and mature ~130kDa (Hoffmann, J., et al. (2000). Eur. J. Cell. Biol.79(12):905-914.). The antibody is known to cross react with APLP2 (Slunt, H.H., et al. (1994). J. Biol. Chem. 269(4):2637-2644).

Immunogen

Epitope: Amino acids 66-81 of APP.
Purified recombinant Alzheimer precursor A4 (pre-A4695) fusion protein.

Application

Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, Alexa Fluor 555 is an antibody against APP A4 for use in Immunocytochemistry.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
The unconjugated antibody (Cat. No. MAB348) is shown to be suitable also for immunohistochemistry (for both frozen and paraffin-embedded sections), immunofluorescence, and Western blotting applications.

Quality

Evaluated by Immunocytochemistry in SH-5YSY cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected APP A4 in SH-5YSY cells.

Target description

~110/120/130 kDa observed.

Physical form

Protein A purified
Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service