Skip to Content
Merck
All Photos(1)

Key Documents

MAB3414

Sigma-Aldrich

Anti-Cytokeratin 8 Antibody, clone 4.1.18

clone 4.1.18, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$785.00

$785.00


Estimated to ship on18 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1688

Request a Bulk Order

Select a Size

Change View
50 μG
$785.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$785.00


Estimated to ship on18 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1688

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4.1.18, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KRT8(3856)

Specificity

The antibody reacts with human cytokeratin 8. There is no cross reactivity with human cytokeratin No. 7, 17, 18, or 19. Anti-cytokeratin No. 8 decorates single epithelia in stomach, colon, and small intestine, gall bladder, liver (bile ducts and hepatocytes) as well as pancreatic ducts and mammary gland ducts.

Immunogen

Cytoskeletal preparation of MCF-7 cells.

Application

This Anti-Cytokeratin 8 Antibody, clone 4.1.18 is validated for use in WB, IC, IH, IH(P) for the detection of Cytokeratin 8.
Western blot Immunocytochemistry Immunohistochemistry: Frozen sections, acetone fixed (5-10 μg/mL)

Paraffin sections, after trypsin treatment (5-20 μg/mL)

Optimal-working dilutions must be determined by end user.

Linkage

Replaces: 04-588

Physical form

Format: Purified
Lyophilized from 0.02 M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristián Vela Hinojosa et al.
PloS one, 8(9), e75062-e75062 (2013-10-03)
The mammary gland has long drawn the attention of the scientific community due to the limited knowledge of some fundamental aspects involved in the control of its function. Myotis velifer, a microchiropteran species, provides an interesting model to study some
Miranda Batchelor et al.
EMBO reports, 5(1), 104-110 (2004-01-08)
While remaining extracellular, enteropathogenic Escherichia coli (EPEC) establish direct links with the cytoskeleton of the target epithelial cell leading to the formation of actin-rich pedestals underneath attached bacteria. The translocated adaptor protein Tir forms the transmembrane bridge between the cytoskeleton
Stanislava Merjava et al.
Investigative ophthalmology & visual science, 52(2), 787-794 (2010-10-12)
The purpose of this study was to investigate the expression of cytokeratin (CK) 8 in the corneoconjunctival epithelium. In 17 cadaveric corneoscleral discs and 3 other discs, the presence of CK8 alone or CK8, together with CK3, CK15, vimentin, and
Leandi Krüger et al.
American journal of physiology. Gastrointestinal and liver physiology, 313(3), G180-G191 (2017-06-03)
Esophageal injury is a risk factor for diseases such as Barrett's esophagus (BE) and esophageal adenocarcinoma. To improve understanding of signaling pathways associated with both normal and abnormal repair, animal models are needed. Traditional rodent models of esophageal repair are
Sinyoung Kim et al.
International journal of molecular medicine, 27(5), 701-706 (2011-02-25)
Although recent studies have demonstrated the in vitro hepatic differentiation potential of mesenchymal stem cells (MSCs), the evidence supporting the in vivo engraftment of MSCs, hepatic differentiation and improvement of hepatic function is still lacking. We investigated in vivo hepatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service