Skip to Content
Merck
All Photos(1)

Key Documents

MAB3301

Sigma-Aldrich

Anti-TIMP-1 Antibody, clone 147-6D11

clone 147-6D11, Chemicon®, from mouse

Synonym(s):

Tissue Inhibitor of Metalloproteinase-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$743.00

$743.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

147-6D11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TIMP1(7076)

Specificity

The antibody specifically reacts with human TIMP-1. This is a purified mouse monoclonal antibody to recombinant human tissue inhibitor of metalloproteinases-1 (recombinant TIMP-1).

SPECIES REACTIVITY:

Does not cross-react with mouse and rat TIMP-1.

Immunogen

Mouse Mac-1

Application

Immunoblotting: Suggested Dilution 1μg/mL

Immunohistochemistry on paraffin-embedded tissue sections using PLP fixation; not tested on traditional formalin fixed tissues. Antigen retrieval is HIER methods

ElA
This Anti-TIMP-1 Antibody, clone 147-6D11 is validated for use in ELISA, WB, IH(P) for the detection of TIMP-1.

Linkage

Replaces: MAB1338

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Tapia-Pizarro et al.
Human reproduction (Oxford, England), 28(8), 2215-2227 (2013-05-23)
Are secreted extracellular matrix (ECM) remodelling elements, relevant to embryo implantation and placentation, modified by hCG in endometrial stromal cells (ESCs)? hCG decreases tissue inhibitor of metalloproteinase 1 (TIMP-1) secretion in ESCs, thereby facilitating extravillous trophoblast invasion in vitro. Successful
Marcia Riboldi et al.
Reproduction (Cambridge, England), 160(5), 673-684 (2020-10-17)
During embryo implantation, endometrial angiogenesis is regulated by signals originating from the endometrium itself and the developing embryo. It has been suggested that hCG may play a pro-angiogenic role; therefore, we sought to understand its regulatory role in blood vessel
Philipp Christian Seppelt et al.
PloS one, 11(2), e0148012-e0148012 (2016-02-04)
Marfan syndrome is an autosomal dominant inherited disorder of connective tissue. The vascular complications of Marfan syndrome have the biggest impact on life expectancy. The aorta of Marfan patients reveals degradation of elastin layers caused by increased proteolytic activity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service