Skip to Content
Merck
All Photos(1)

Key Documents

MAB3140

Sigma-Aldrich

Anti-Na+/H+ Exchanger-1 Antibody, CT, clone 4E9

clone 4E9, Chemicon®, from mouse

Synonym(s):

NHE1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$785.00

$785.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$785.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$785.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E9, monoclonal

species reactivity

amphibian, fish, avian, vertebrates (including including fish, amphibians, birds and mammals)

species reactivity (predicted by homology)

mouse, mammals, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG1

suitability

not suitable for immunoprecipitation

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC9A1(6548)

General description

Mammalian NHE (Na+/H+ Exchanger) is an integral membrane protein that is known to regulate intracellular pH by removing a proton in exchange for an extracellular sodium ion. There are nine known isofoms of NHE. The best characterized is NHE1, involved not only in intracellular pH control, but cell-volume control, cytoskeletal organization, heart disease and cancer. Transport by the NHE plays a pivotal role in the damage caused to the human myocardium during and following a myocardial infarction, and it is considered to represent a key step in the oncogenic transformation of cancerous cells. NHE comprises an N-terminal membrane domain that transports ions, and a C-terminal cytoplasmic domain that regulates activity and mediates cytoskeletal interactions. NHE2 - NHE5 isoforms are localized to the plasma membrane but exhibit greater tissue-specificity. NH2 and NH3 are more highly expressed in the kidney and intestine. Structurally, a single transmembrane segment from NHE1 has been solved, and the structure of bacterial Na+/H+ antiporter NhaA has been elucidated.

Specificity

Na+/H+ exchanger, isoform NHE1

Immunogen

Epitope: C-terminus
MBP fusion protein containing the entire C-terminal, hydrophylic domain of porcine NHE1.

Application

Anti-Na+/H+ Exchanger-1 Antibody, C-terminus, clone 4E9 detects level of Na+/H+ Exchanger-1 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Works poorly for immunohistochemistry and immunocytochemistry.
Not recommended for immunoprecipitation.

Immunoblotting:
1:1,000 dilution used on a previous lot.

Optimal working dilutions must be determined by the user.

Quality

Routinely evaluated by Western Blot on Human Kidney lysates.

Western Blot:
1:500 dilution of this lot detected Na+/H+ Exchanger-1 on 10 μg of Human Kidney lysates.

Target description

~100-110 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1 liquid in buffer containing 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Kidney tissue (basolateral membrane)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Juel et al.
The Journal of physiology, 548(Pt 2), 639-648 (2003-03-04)
Chronic hypoxia is accompanied by changes in blood and skeletal muscle acid-base control. We hypothesized that the underlying mechanisms include altered protein expression of transport systems and the enzymes involved in lactate, HCO3- and H+ fluxes in skeletal muscle and
Sebastiano Vilella et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 13(4), 207-214 (2003-07-24)
In thyroid cells, the intracellular pH plays a key role in the control of the iodide uptake, since iodide accumulation is associated with an intracellular acidification. In the present paper we studied the kinetic proprieties of the Na(+)/H(+) antiporter (NHE)
Lionel Blanc et al.
American journal of hematology, 90(3), 235-241 (2014-12-18)
Genetic ablation of the ferrireductase STEAP3, also known as TSAP6, leads to severe microcytic and hypochromic red cells with moderate anemia in the mouse. However, the mechanism leading to anemia is poorly understood. Previous results indicate that TSAP6/Steap3 is a
Cian McGinley et al.
Experimental physiology, 101(12), 1565-1580 (2016-10-01)
What is the central question of this study? Following a training intervention, how is the interpretation of adaptations in skeletal muscle H+ transporters influenced by biopsy timing in the context of individual protein and mRNA kinetics after the final exercise
Laurent A Messonnier et al.
Nutrients, 14(3) (2022-03-13)
It remains unclear whether sickle cell trait (SCT) should be considered a risk factor during intense physical activity. By triggering the polymerization-sickling-vaso-occlusion cascade, lactate accumulation-associated acidosis in response to high-intensity exercise is believed to be one of the causes of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service