Skip to Content
Merck
All Photos(6)

Key Documents

MAB2258

Sigma-Aldrich

Anti-Syntaxin-3 Antibody, clone 1-146

clone 1-146, from mouse

Synonym(s):

syntaxin 3, syntaxin 3A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$806.00

$806.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$806.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$806.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1-146, monoclonal

species reactivity

human, rat

species reactivity (predicted by homology)

mouse

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STX3(6809)
rat ... Stx3(81802)

General description

Syntaxin-3, also known as Stx3, is a member of the SNARE family which are complexes involved in membrane fusion. Syntaxin-3 is expressed in epithelial cells and is found apically in MDCK and Caco-2 cells (1).

Specificity

This antibody recognizes Syntaxin-3.

Immunogen

GST-tagged recombinant protein corresponding to residues contained within rat Syntaxin-3.

Application

Detect Syntaxin-3 using this Anti-Syntaxin-3 Antibody, clone 1-146 validated for use in WB, IP, IH.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Quality

Evaluated by Western Blot in rat brain tissue lysate.
Western Blot Analysis: : 0.5 µg/mL dilution of this antibody detected Syntaxin-3 on 15 µg of rat brain tissue lysate.

Target description

Approx. 33 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in buffer containing 0.1 M Tris-glycine, pH 7.4, 150 mM NaCl with 0.05% NaN3.

Storage and Stability

Maintain refrigerated at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian J Giovannone et al.
The Journal of biological chemistry, 293(15), 5478-5491 (2018-02-25)
Syntaxins are a conserved family of SNARE proteins and contain C-terminal transmembrane anchors required for their membrane fusion activity. Here we show that Stx3 (syntaxin 3) unexpectedly also functions as a nuclear regulator of gene expression. We found that alternative
Daniëlle Rianne José Verboogen et al.
eLife, 6 (2017-05-20)
SNARE proteins play a crucial role in intracellular trafficking by catalyzing membrane fusion, but assigning SNAREs to specific intracellular transport routes is challenging with current techniques. We developed a novel Förster resonance energy transfer-fluorescence lifetime imaging microscopy (FRET-FLIM)-based technique allowing
Ritika Chatterjee et al.
Traffic (Copenhagen, Denmark), 24(7), 270-283 (2023-04-28)
Intracellular membrane fusion is mediated by membrane-bridging complexes of soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNAREs). SNARE proteins are one of the key players in vesicular transport. Several reports shed light on intracellular bacteria modulating host SNARE machinery to establish
Ying Hsu et al.
Molecular therapy. Nucleic acids, 31, 164-181 (2023-01-27)
Blindness in Bardet-Biedl syndrome (BBS) is caused by dysfunction and loss of photoreceptor cells in the retina. BBS10, mutations of which account for approximately 21% of all BBS cases, encodes a chaperonin protein indispensable for the assembly of the BBSome
Adrian J Giovannone et al.
Molecular biology of the cell, 28(21), 2843-2853 (2017-08-18)
Syntaxin 3 (Stx3), a SNARE protein located and functioning at the apical plasma membrane of epithelial cells, is required for epithelial polarity. A fraction of Stx3 is localized to late endosomes/lysosomes, although how it traffics there and its function in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service