Skip to Content
Merck
All Photos(2)

Key Documents

MAB2252Z-I

Sigma-Aldrich

Anti-Integrin β1 Antibody, clone N29 (Azide Free)

clone N29, Chemicon®, from mouse

Synonym(s):

CD29, MAB2252

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$682.00

$682.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1271

Request a Bulk Order

Select a Size

Change View
100 μG
$682.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$682.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1271

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

N29, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgGκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Related Categories

Specificity

Reacts with the human integrin beta1 subunit. Specificity verified by preclearing of beta1in immunoprecipitation, flow cytometry on transfectant cells displaying human beta1 integrin, and reactivity with purified beta1. Recognizes an epitope cluster distinct from MAB2250 and MAB2251(Wilkins et al., 1996).

Immunogen

Jurkat T-leukemic cell line

Application

Immunohistochemistry (Paraffin) Analysis: A 1:50-2,000 dilution from a representative lot detected Integrin β1 in the membrane of tubule epithelial cells and cells of the glomeruli in human kidney tissue, membrane of alveolar cells, endothelial cells, and bronchiole epithelial cells of human lung tissue.

Immunoprecipitation: A representative lot of this antibody clone was used in immunoprecipitation.

Immunohistochemistry: A representative lot of this antibody clone was used in immunohistochemistry on frozen sections.

Flow Cytometry: A representative lot of this antibody clone was used in flow cytometry.

Functional Activity Assay: A representative lot of this antibody clone was used to stimulate adhesion of cells to extracellular matrix proteins (Wilkins, J.A. et al., 1996).

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin β1 Antibody, clone N29 (Azide Free) is validated for use in Flow Cytometry and Immunohistochemistry and Immunoprecipitation and Affects Function and Cell Culture and Western Blotting for the detection of Integrin β1.

Quality

Western Blot Analysis: 0.5 µg/mL of the antibody detected Integrin β1 in 10 µg of U251 cell lysate.

Target description

88 kDa

Physical form

Format: Purified
Protein A purified
Purified cultured supernatant in PBS without preservatives.

Storage and Stability

Store at -20°C in undiluted aliquots.
Avoid repeated freeze-thaw cycles.

Analysis Note

Control
Tonsil, human skin, human kidney tissue

U251 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rania Hadjisavva et al.
Cell reports, 40(3), 111091-111091 (2022-07-21)
Cadherins and integrins are intrinsically linked through the actin cytoskeleton and are largely responsible for the mechanical integrity and organization of tissues. We show that cadherin clustering stimulates and spatially guides integrin activation. Adherens junction (AJ)-associated integrin activation depends on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service