Skip to Content
Merck
All Photos(1)

Key Documents

MAB2137

Sigma-Aldrich

Anti-Homing Receptor Antibody, clone A020

clone A020, Chemicon®, from rat

Synonym(s):

CD44s, HCAM, Pgp-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$825.00

$825.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$825.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$825.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

A020, monoclonal

species reactivity

mouse, human, rat, pig, rabbit, canine, bovine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CD44(960)

Related Categories

Specificity

Antibody reacts with the standard CD44 isoform (CD44s) that lacks exon sequences v1-v10. Also called human CD44 (85-95 kDa) gp90{Hermes} and mouse gp85 (Pgp-1).

Immunogen

Purified human blood lymphocyte CD44

Application

Anti-Homing Receptor Antibody, clone A020 is an antibody against Homing Receptor for use in ELISA, FC, IC, IH & WB.
ELISA 1:2000

Flow Cytometry :1:1000

Immunofluorescence 1:200

Immunohistostaining of paraffin embedded tissues. 1:250; antigen retrieval is usually not needed.

Immunoprecipitation 1:200

Western blotting: non-reduced conditions only; 90kDa band is detected; blocking PBS/5% milk protein, primary antibody dilution 1:1000 o/n incubation.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Target description

90 kDa

Physical form

Format: Purified
Liquid in 10 mM sodium phosphate, pH 7.5 with 150 mM NaCl and 0.01% sodium azide.
Protein A purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Cells of epithelium and highly expressed by carcinomas

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameerah Shaheen et al.
Stem cell reviews and reports, 12(4), 492-499 (2016-05-22)
Colorectal cancers (CRCs) form a disorganized hierarchy of heterogeneous cell populations on which current chemotherapy regimens fail to exert their distinctive cytotoxicity. A small sub-population of poorly differentiated cancer stem-like cells (CSCs), also known as cancer initiating cells, may exhibit
Laura Novellasdemunt et al.
Stem cell reports, 18(2), 570-584 (2023-01-21)
Adenomatous polyposis coli (APC) mutation is the hallmark of colorectal cancer (CRC), resulting in constitutive WNT activation. Despite decades of research, targeting WNT signaling in cancer remains challenging due to its on-target toxicity. We have previously shown that the deubiquitinating
Adam C Midgley et al.
The Journal of biological chemistry, 290(18), 11218-11234 (2015-02-27)
Hyaluronan (HA) promotes transforming growth factor (TGF)-β1-driven myofibroblast phenotype. However, HA can also have disease-limiting activity. Bone morphogenetic protein-7 (BMP7) is an antifibrotic cytokine that antagonizes TGF-β1, and isolated studies have demonstrated that HA can both mediate and modulate BMP7
Carmela Ricciardelli et al.
BMC cancer, 13, 476-476 (2013-10-16)
Hyaluronan (HA) an important component of the extracellular matrix, has been linked to tumor progression and drug resistance in several malignancies. However, limited data is available for ovarian cancer. This study investigated the role of hyaluronan (HA) and a potential
Hsiao-Han Wang et al.
American journal of translational research, 9(11), 4785-4806 (2017-12-09)
Glioblastoma multiforme (GBM) is one of the most malignant and aggressive brain tumors with great amount of hyaluronan (HA) secretion and CD44 overexpression (HA receptor). CD44 has been suggested as a cancer stem cells (CSCs) marker. However, several clinical studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service