Skip to Content
Merck
All Photos(2)

Key Documents

MAB2059Z

Sigma-Aldrich

Anti-Integrin β4 Antibody, clone ASC-8 (Azide Free)

clone ASC-8, Chemicon®, from mouse

Synonym(s):

CD104, MAB2059

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$635.00

$635.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$635.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$635.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ASC-8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB4(3691)

Related Categories

Application

Anti-Integrin β4 Antibody, clone ASC-8 (Azide Free) is an antibody against Integrin β4 for use in FC, IP, IH, FUNC.

Quality

Flow Cytometry Analysis:
0.5 µg of the antibody was used to detect Integrin β4 in 1x10^6 A431 cells.
0.5 µg of the antibody was used to detect Integrin β4 in 1x10^6 HeLa cells.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Rath et al.
Cellular and molecular life sciences : CMLS, 79(6), 340-340 (2022-06-07)
Cerebral cavernous malformations (CCM) are low-flow vascular lesions prone to cause severe hemorrhage-associated neurological complications. Pathogenic germline variants in CCM1, CCM2, or CCM3 can be identified in nearly 100% of CCM patients with a positive family history. In line with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service