Skip to Content
Merck
All Photos(1)

Key Documents

MAB1976B

Sigma-Aldrich

Anti-Integrin αVβ3 Antibody, clone LM609, biotin conjugated

clone LM609, Chemicon®, from mouse

Synonym(s):

Vitronectin Receptor, CD51/CD61

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$793.00

$793.00


Estimated to ship on29 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$793.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$793.00


Estimated to ship on29 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

LM609, monoclonal

species reactivity

canine, chicken, pig, avian, human, bovine, rabbit, monkey

should not react with

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Related Categories

General description

The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence, known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor (Cheresh, 1991). The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented. The adhesion receptor, integrin alphaVbeta3, appears to be selectively expressed on growing blood vessels and has been identified as a marker of angiogenic vascular tissue (Brooks, 1994). Due to its involvement in angiogenesis, integrin alphaVbeta3 is one of the most intensely studied of the integrin receptors.

Specificity

Reacts with the vitronectin receptor alphaVbeta3 complex, an RGD-directed adhesion receptor. LM609 has been demonstrated to block adhesion of a human melanoma cell line (M21) to vitronectin, fibrinogen and von Willebrand factor, as well as to a synthetic RGD containing peptide (Cheresh, 1987). In chick chorioallentoic membranes, LM609 was shown to block angiogenesis induced by bFGF and TNFalpha but had no effect on pre-existing vessels (Brooks, 1994). While LM609 does block cell attachment to RGD containing ligands, it does not interact directly with the RGD binding site. Instead, LM609 appears to be an allosteric inhibitor of integrin alphaVbeta3, which binds to a conformational epitope resulting from the post-translational association of the alphaV and beta3 subunits.

Immunogen

Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, 1987).

Application

Detect Integrin αVβ3 using this Anti-Integrin αVβ3 Antibody, clone LM609, biotin conjugated validated for use in FC, IH & IP.
Immunoprecipitation: specifically immunoprecipitates the integrin alphaVbeta3 complex which, when resolved by SDS-PAGE under reducing conditions, results in an approximately 130 kDa band corresponding to the alphaV subunit and a 105 kDa band corresponding to the beta3 subunit (Cheresh, 1987).

Immunofluorescence: 5-10 μg/mL for staining of bFGF-treated 6 mm cryosections of chick chorioallantoic membrane fixed with acetone (Brooks, 1994). LM609 is also effective on 4% paraformaldehyde fixed, frozen tissues and cells. However it is not effective for immunohistochemical staining of paraffin-embedded tissue sections.

Inhibits adhesion of cells to vitronectin coated surfaces at 10-25 μg/mL.

Optimal working dilutions and protocols must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Liquid in 0.01M PBS (pH 7.1), 0.1% sodium azide with 15 mg/mL BSA added as a stabilizer.
Protein A purified

Storage and Stability

Maintain at 2–8°C in undiluted aliquots for up to 6 months.

Analysis Note

Control
POSITIVE CONTROL: Human myeloma cell lines M21 and MoalphaV (Chen, 1995).

NEGATIVE CONTROL: Mo cells (this cell line was derived from M21 but does not express integrin alphaV

Chen, 1995).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaoping Qin et al.
Journal of cell communication and signaling, 17(2), 287-296 (2023-05-28)
Skin primarily comprises a collagen-rich extracellular matrix (ECM) that provides structural and functional support to the skin. Aging causes progressive loss and fragmentation of dermal collagen fibrils, leading to thin and weakened skin (Dermal aging). We previously reported that CCN1
Daphne Lobeek et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 61(2), 270-275 (2019-09-15)
Arteriovenous malformations (AVMs) have an inherent capacity to form new blood vessels, resulting in excessive lesion growth, and this process is further triggered by the release of angiogenic factors. 68Ga-labeled arginine-glycine-aspartate tripeptide sequence (RGD) PET/CT imaging may provide insight into
Roland Haubner et al.
PLoS medicine, 2(3), e70-e70 (2005-03-24)
The integrin alphavbeta3 plays an important role in angiogenesis and tumor cell metastasis, and is currently being evaluated as a target for new therapeutic approaches. Several techniques are being studied to enable noninvasive determination of alphavbeta3 expression. We developed [(18)F]Galacto-RGD
Nadia Withofs et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 56(3), 361-364 (2015-02-07)
This study aimed to correlate (18)F-FB-mini-PEG-E[c(RGDyK)](2) ((18)F-FPRGD2) uptake to integrin αvβ3 expression and angiogenesis in renal tumors. (18)F-FPRGD2 PET/CT was performed on 27 patients before surgical resection (median 4 d) of a renal mass. The (18)F-FPRGD2 uptake was compared with
Qi Sun et al.
NPJ breast cancer, 8(1), 5-5 (2022-01-15)
Breast cancer cells with stem-like properties are critical for tumor progression, yet much about these cells remains unknown. Here, we characterize a population of stem-like breast cancer cells expressing the integrin αvβ3 as transcriptionally related to activated stem/basal cells in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service