Skip to Content
Merck
All Photos(2)

Key Documents

MAB1976-AF555

Sigma-Aldrich

Anti-Integrin αVβ3 Antibody, clone LM609, Alexa Fluor 555 Conjugate

clone LM609, from mouse, ALEXA FLUOR 555

Synonym(s):

Integrin alpha-V, Vitronectin receptor subunit alpha, CD51, Integrin alpha-V heavy chain, Integrin alpha-V light chain, Integrin αVβ3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$793.00

$793.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$793.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$793.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

purified antibody

antibody product type

primary antibodies

clone

LM609, monoclonal

species reactivity

chicken, canine, monkey, rabbit, pig, human, avian, bovine

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

Integrin alpha-V (CD51, Vitronectin receptor subunit alpha; UniProt P06756) and Integrin beta-3 (CD61, GPIIIa, Platelet membrane glycoprotein IIIa; UniProt P05106) are encoded by the ITGAV (MSK8, VNRA; Gene ID 3685) and ITGB3 (GP3A; Gene ID 3690) gene, respectively, in human. The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and a beta subunit that associate in a non-covalent manner. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence, known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor. The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented. The adhesion receptor, integrin alphaVbeta3, appears to be selectively expressed on growing blood vessels and has been identified as a marker of angiogenic vascular tissue. Due to its involvement in angiogenesis, integrin alphaVbeta3 is among of the most studied integrin receptors.

Specificity

Reacts with the vitronectin receptor alphaVbeta3 complex, an RGD-directed adhesion receptor. LM609 has been demonstrated to block adhesion of a human melanoma cell line (M21) to vitronectin, fibrinogen and von Willebrand factor, as well as to a synthetic RGD containing peptide (Cheresh, D.A., and Spiro R.C. (1987). J Biol Chem. 262(36):17703-11; Cheresh, D.A. (1987). Proc. Natl. Acad. Sci. U.S.A. 84(18):6471-6475). In chick chorioallentoic membranes, LM609 was shown to block angiogenesis induced by bFGF and TNFalpha but had no effect on pre-existing vessels (Brooks, P.C., et al. (1994). Science. 264(5158):569-571). While LM609 does block cell attachment to RGD containing ligands, it does not interact directly with the RGD binding site. Instead, LM609 appears to be an allosteric inhibitor of integrin alphaVbeta3, which binds to a conformational epitope resulting from the post-translational association of the alphaV and beta3 subunits.

Immunogen

Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, D.A., and Spiro R.C. (1987). J. Biol. Chem. 262(36):17703-11).

Application

Anti-Integrin αVβ3 Antibody, clone LM609, Alexa Fluor 555 Conjugate is an antibody against Integrin αVβ3 for use in Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Integrin αVβ3 in HeLa cells.
The unconjugated antibody (Cat. No. MAB1976) is shown to be suitable also for functional assay, flow cytometry, immunofluorescence, immunohistochemistry, and immunoprecipitation applications.

Quality

Evaluated by Immunocytochemistry in HepG2 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Integrin αVβ3 in HepG2 cells.

Target description

~116 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service