Skip to Content
Merck
All Photos(1)

Key Documents

MAB1951

Sigma-Aldrich

Anti-Integrin beta 1 (CD29) Antibody

CHEMICON®, mouse monoclonal, P4G11

Synonym(s):

CD29

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$785.00

$785.00


Estimated to ship on31 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1230

Request a Bulk Order

Select a Size

Change View
100 μG
$785.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$785.00


Estimated to ship on31 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1230

Request a Bulk Order

Product Name

Anti-Integrin β1 Antibody, clone P4G11, clone P4G11, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P4G11, monoclonal

species reactivity

monkey, rabbit, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
cell culture | mammalian: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Related Categories

Specificity

Reacts with calcium-dependent epitopes on the integrin β1 subunit. When bound to β1, P4G11, activates β1 dependent binding to extra cellular matrix substrates.

Immunogen

Human IL-1beta activated endothelial cells

Application

Anti-Integrin β1 Antibody, clone P4G11 is an antibody against Integrin β1 for use in ELISA, FC, IC, IH, IH(P), IP, FUNC, CULT.
Immunohistochemistry: for use on acetone or mild paraformaldehyde fixed tissue (2-2.5% PFA, short time) frozen and 4% PFA paraffin embedded {Qin et al 2003 1:500, citrate HEIR treated}

Ca++ required for binding. Ca++ must be used in blocking, washing, and antibody incubation buffers.

Traditional formalin fixation is not recommended.

Immunocytochemistry: 2 ug/mL (for use on numerous cell lines as long as Ca2+ is present)

Immunoprecipitation: 5 μg/400 μL lysate (0.5% triton PBS plus Ca++)

FACS: 2-5μg per 10e6 cells, in the presence of Ca++.

Stimulation of cells: 2-10 μg/ml per 10e6 cells. Binding buffer must contain 1mM Ca++.

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Protein A Purified immunoglobulin in 0.02M PBS, 0.25M NaCl, pH 7.6, with 0.1% NaN3.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Tonsil, human skin tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna M Joy et al.
Journal of cell science, 116(Pt 21), 4409-4417 (2003-09-18)
Glioma cells that migrate out of the main tumor mass into normal brain tissue contribute to the failure of most gliomas to respond to treatment. Treatments that target migratory glioma cells may enhance the therapeutic response. Multiple lines of evidence
Jesus Vega-Lugo et al.
The Journal of cell biology, 221(7) (2022-05-14)
Colocalization analysis of multicolor microscopy images is a cornerstone approach in cell biology. It provides information on the localization of molecules within subcellular compartments and allows the interrogation of known molecular interactions in their cellular context. However, almost all colocalization
Convergence of eicosanoid and integrin biology: 12-lipoxygenase seeks a partner.
Tang, K; Cai, Y; Joshi, S; Tovar, E; Tucker, SC; Maddipati, KR; Crissman, JD; Repaskey, WT; Honn, KV
Molecular Cancer null
C Radel et al.
American journal of physiology. Heart and circulatory physiology, 288(2), H936-H945 (2004-10-09)
To identify the role of caveolin-1 in integrin mechanotransduction, we exposed bovine aortic endothelial cells to 10 dyn/cm2 of laminar shear stress. Caveolin-1 was acutely and transiently phosphorylated with shear, occurring downstream of beta1-integrin activation as the beta1-integrin blocking antibody
Francesca Ricciardiello et al.
Cell death & disease, 9(3), 377-377 (2018-03-09)
Cancer aberrant N- and O-linked protein glycosylation, frequently resulting from an augmented flux through the Hexosamine Biosynthetic Pathway (HBP), play different roles in tumor progression. However, the low specificity and toxicity of the existing HBP inhibitors prevented their use for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service