Skip to Content
Merck
All Photos(2)

Key Documents

MAB1904

Sigma-Aldrich

Anti-Laminin-1 A&B chains Antibody

CHEMICON®, rat monoclonal, AL-2

Synonym(s):

laminin alpha1, laminin beta1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$651.00

$651.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$651.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$651.00


Estimated to ship on02 June 2025


Request a Bulk Order

Product Name

Anti-Laminin-1 A&B chains Antibody, cross region, clone AL-2, clone AL-2, Chemicon®, from rat

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AL-2, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lama1(16772)

General description

Laminins are secreted glycoproteins incorporated into cell-associated extracellular matrices and an integral part of the structural scaffolding in most animal tissues. The laminin protein is made up of three polypeptide chains; α, β and γ, accounting for its flexibility in connecting various molecules. When originally characterized, laminin-1 clone AL-2 reacted with Engelbreth-Holm-Swarm (EHS) mouse laminin A & B chains on the long arm near the cross region, (Skubitz, 1987). More recently, investigations, (Schèele, 2006) have shown that the clone binds laminin β1 under reducing conditions as well as native ELISAs and that the affinity is increased by the presence of the laminin α1 chain. Thus there is significant laminin β1 reactivity with clone AL-2.

Specificity

Other species have not been tested.
Reacts with EHS mouse laminin A & B chains on the long arm near the cross region.

Immunogen

Epitope: Near cross region
Laminin isolated from EHS mouse sarcoma.

Application

Detect Laminin-1 A&B chains using this Anti-Laminin-1 A&B chains Antibody, cross region, clone AL-2 validated for use in ELISA, IF, IH(P), IP & WB.
Immunofluorescence:
An independent laboratory (Schèele, S., 2006) has verified anti-laminin clone AL-2 to work using immunofluorescence techniques on mouse muscle tissue, including both transgenic and wild-type tissues.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated on a representative lot by Western blot on purified mouse laminin-1.

Target description

~200 kDa

Physical form

Format: Purified
Protein G Purified
Purified in 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% NaN3.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George S Karagiannis et al.
Molecular oncology, 8(2), 178-195 (2013-11-26)
It has been suggested that cancer-associated fibroblasts (CAFs) positioned at the desmoplastic areas of various types of cancer are capable of executing a migratory program, characterized by accelerated motility and collective configuration. Since CAFs are reprogrammed derivatives of normal progenitors
H Kurata et al.
The Journal of allergy and clinical immunology, 103(5 Pt 2), S471-S484 (1999-05-18)
Adenoviruses have several specific features useful for gene therapy. They infect various lineages of cells irrespective of cell cycle status. However, the exact mechanism of their infection and in vivo kinetics as a gene expression vector have not been elucidated.
Ghaidaa Kashgari et al.
Developmental cell, 52(6), 764-778 (2020-02-29)
The fusion of digits or toes, syndactyly, can be part of complex syndromes, including van der Woude syndrome. A subset of van der Woude cases is caused by dominant-negative mutations in the epithelial transcription factor Grainyhead like-3 (GRHL3), and Grhl3-/-mice
Jangwook P Jung et al.
Biomacromolecules, 14(9), 3102-3111 (2013-07-24)
Limiting the precise study of the biochemical impact of whole molecule extracellular matrix (ECM) proteins on stem cell differentiation is the lack of 3D in vitro models that can accommodate many different types of ECM. Here we sought to generate
Raven Diacou et al.
Cell reports, 25(9), 2510-2523 (2018-11-30)
Gene regulation of multipotent neuroretinal progenitors is partially understood. Through characterizing Six3 and Six6 double knockout retinas (DKOs), we demonstrate Six3 and Six6 are jointly required for the maintenance of multipotent neuroretinal progenitors. Phenotypes in DKOs were not found in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service