Skip to Content
Merck
All Photos(1)

Key Documents

MAB1233

Sigma-Aldrich

Anti-Estradiol Antibody, clone 6E1

ascites fluid, clone 6E1, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$524.00

$524.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μL
$524.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$524.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6E1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ESR1(2099)

Specificity

Estradiol-6-BSA

Application

Detect Estradiol with Anti-Estradiol Antibody, clone 6E1 (Mouse Monoclonal Antibody), that has been shown to work in ELISA.
Immunoassay.

Final working dilutions must be determined by end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Linkage

Replaces: CBL69

Storage and Stability

Maintain at -20°C in convenient aliquots for up to 12 months. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service