Skip to Content
Merck
All Photos(1)

Key Documents

MAB1202

Sigma-Aldrich

Anti-CD42a Antibody, platelet lb-IX, clone FMC-25

clone FMC-25, Chemicon®, from mouse

Synonym(s):

Anti-CD42a, Anti-GPIX

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$558.00

$558.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
$558.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$558.00


Available to ship on31 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FMC-25, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GP9(2815)

Specificity

This antibody belongs to CD42a (assigned at the Fourth International Workshop on Leukocyte Differentiation Antigens, Vienna, 1989) (Knapp, 1989), and binds to the 22,000 dalton IX subunit of the glycoprotein Ib-IX complex (Zola et al., 1984; Berndt et al., 1985a,b). FMC-25 failed to bind to platelets from patients with Bernard-Soulier Syndrome (Berndt et al., 1983). The antibody is specific for cells of the platelet megakaryocyte lineage. It reacts with a proportion of acute myeloid leukemias being useful in identifying those of the megakaryoblastic subtype (San Miguel et al., 1985, 1986).

Cell reactivity:

The antibody is specific for cells of the platelet megakaryocyte lineage. It reacts with a proportion of acute myeloid leukemias being useful in identifying those of the megakaryoblastic subtype(San Miguel et al., 1985, 1986).

Immunogen

Epitope: platelet lbalpha complex
Peripheral blood mononuclear cells isolated on Ficoll® hypaque.

Application

Anti-CD42a Antibody, platelet lb-IX, clone FMC-25 detects level of CD42a & has been published & validated for use in FC, IC, IH.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
The glycoprotein Ib-IX complex on the surface of normal platelets is a heterotrimer of Ib-alpha, Ib-beta and IX subunits (Berndt, 1989). Ib-IX complex is an essential component of platelet adhesion to vascular subendothelium and is also important in the maintenance of platelet shape (Berndt et al., 1988).

This antibody is suitable for enumeration of platelets by flow cytometry and immunoperoxidase staining of frozen tissue sections or air dried smear preparations (San Miguel et al., 1986), and for identification of megakaryoblasts in acute myeloid leukemia or acute myelofibrosis (San Miguel et al., 1985; San Miguel et al., 1986).

SUGGESTED USAGE DILUTION

1. Flow cytometry and indirect immunofluorescence 1:25

Dilute with isotonic buffer. Use 50 μl of diluted antibody per 1 x 10E6 platelets in 100 μl of buffer.

2. Indirect immunoperoxidase staining - the final dilution will depend on the assay conditions and detection system employed. However, a dilution of at least 1:20 will be applicable to most commonly used systems.

Physical form

Format: Purified
Purified immunoglobulin. The antibody is supplied in 25mM Tris-HCl, 0.4M NaCl, pH 8.0 containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Storage and Stability

Store at 2 to 8°C, for up to 6 months. For prolonged periods, store below -20°C in undiluted aliquots. AVOID REPEATED FREEZE/THAW CYCLES.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Ficoll is a registered trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Yan et al.
Cell death & disease, 12(11), 955-955 (2021-10-18)
Platelets are generated from the cytoplasm of megakaryocytes (MKs) via actin cytoskeleton reorganization. Zyxin is a focal adhesion protein and wildly expressed in eukaryotes to regulate actin remodeling. Zyxin is upregulated during megakaryocytic differentiation; however, the role of zyxin in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service