Skip to Content
Merck
All Photos(1)

Key Documents

MAB1044

Sigma-Aldrich

Anti-Interleukin-8 Antibody, clone 8M6

clone 8M6, Chemicon®, from mouse

Synonym(s):

IL-8

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$673.00

$673.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
250 μG
$673.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$673.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8M6, monoclonal

species reactivity

bovine, goat, sheep

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
neutralization: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Cxcl8(280828)

Specificity

Reacts with ovine IL-8. No reactivity detected to recombinant ovine IL-1beta, IL-6, MCP-1 or TNFalpha.

Immunogen

Recombinant ovine IL-8

Application

Capture antibody in ELISA. Recommended coating concentration 5 μg/mL. See suggested protocol.

Flow cytometry: use 1 μg/test

Neutralizes recombinant ovine IL-8.

Western blotting

Optimal working dilutions must be determined by the end user.

Suggested ELISA Protocol For Ovine IL-8

1. Coat high capacity microtitre wells (e.g. Nunc maxisorp) with monoclonal anti-IL-8 antibody (Chemicon Catalog Number MAB1044) at a concentration of 5 μg/mL in 0.1 M carbonate buffer pH 9.6 overnight at 4ºC.

2. Wash wells 3 x with PBS + 0.05% Tween 20 (PBST).

3. Add samples to wells. Incubate at room temperature in a humidified box for 1 hour. Wash 3 x with PBST.

4. Dilute rabbit anti-IL-8 (Chemicon Catalog Number AB1840) 1:500 in PBST and add 100 μL to wells. Incubate at room temperature for 30 minutes. Wash 3 x with PBST.

5. Dilute anti-rabbit IgG-HRP to appropriate level in PBST and add 100 μL to wells. Incubate 30 minutes at room temperature. Wash 3 x with PBST.

6. Add 100 μL of TMB chromagen to each well. Incubate for 5-10 minutes and stop reaction with 50 μL of 2M sulfuric acid.

Notes:

The anti-rabbit IgG HRP must be adsorbed against mouse immunoglobulin, and should be checked for non-reactivity with coated plates.

Non-specific binding may be reduced by addition of protein such as skim milk powder to the diluents.

Assay sensitivity may be altered by adjusting incubation times and reagent concentrations.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
This Anti-Interleukin-8 Antibody, clone 8M6 is validated for use in ELISA, FC, NEUT, WB for the detection of Interleukin-8.

Physical form

Format: Purified
Liquid in PBS containing 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Zhu et al.
Biology of reproduction, 104(2), 317-324 (2020-12-11)
High unintended pregnancy rates are partially due to lack of effective nonhormonal contraceptives; development of safe, effective topical vaginal methods will address this need. Preclinical product safety and efficacy assessment requires in vivo testing in appropriate models. The sheep is
Charlotte van Gorp et al.
Nutrients, 11(5) (2019-05-01)
Chorioamnionitis, clinically most frequently associated with Ureaplasma, is linked to intestinal inflammation and subsequent gut injury. No treatment is available to prevent chorioamnionitis-driven adverse intestinal outcomes. Evidence is increasing that plant sterols possess immune-modulatory properties. Therefore, we investigated the potential
Monique G M Willems et al.
American journal of physiology. Lung cellular and molecular physiology, 310(1), L1-L7 (2015-11-01)
Chorioamnionitis, an inflammatory reaction of the fetal membranes to microbes, is an important cause of preterm birth and associated with inflammation-driven lung injury. However, inflammation in utero overcomes immaturity of the premature lung by inducing surfactant lipids and lung gas
Ruth Gussenhoven et al.
Journal of neuroinflammation, 15(1), 113-113 (2018-04-21)
Antenatal infection (i.e., chorioamnionitis) is an important risk factor for adverse neurodevelopmental outcomes after preterm birth. Destructive and developmental disturbances of the white matter are hallmarks of preterm brain injury. Understanding the temporal effects of antenatal infection in relation to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service