Skip to Content
Merck
All Photos(1)

Key Documents

MAB045X

Sigma-Aldrich

Anti-Fluorescein Antibody, clone 5D6.2, Alexa Fluor 488 conjugated

clone 5D6.2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$846.00

$846.00


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$846.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$846.00


Estimated to ship on06 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D6.2, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Fluorescein.

Immunogen

Fluorescein-KLH

Application

Detect Fluorescein using this Anti-Fluorescein Antibody, clone 5D6.2, Alexa Fluor 488 conjugated validated for use in IF.
Optimal working dilutions must be determined by end user.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Physical form

Purified immunoglobulin conjugated to Alexa Fluor 488. Liquid in Phosphate buffer with 15 mg/mL BSA as a stabilizer and 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in the dark for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sydney Aten et al.
Learning & memory (Cold Spring Harbor, N.Y.), 25(5), 214-229 (2018-04-18)
The microRNA miR-132 serves as a key regulator of a wide range of plasticity-associated processes in the central nervous system. Interestingly, miR-132 expression has also been shown to be under the control of the circadian timing system. This finding, coupled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service