Skip to Content
Merck
All Photos(1)

Key Documents

MAB012

Sigma-Aldrich

Anti-Apolipoprotein B Antibody, clone B1

clone B1, Chemicon®, from mouse

Synonym(s):

ApoB

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$694.00

$694.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
$694.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$694.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

B1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
radioimmunoassay: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APOB(338)

Specificity

Recognizes Apolipoprotein B. No cross-reactivity to Apo A-I or Apo A-II.

Application

Competitive RIA

ELISA

Optimal working dilution must be determined by the end user.
This Anti-Apolipoprotein B Antibody, clone B1 is validated for use in ELISA, RIA for the detection of Apolipoprotein B.

Physical form

Format: Purified
Liquid, in 0.015 M potassium phosphate buffer with 0.15M NaCl, 0.1% NaN3, pH 7.2.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Auclair et al.
Scientific reports, 9(1), 17975-17975 (2019-12-01)
Acylated ghrelin (AG) is a gastrointestinal (GI) peptide mainly secreted by the stomach that promotes cytosolic lipid droplets (CLD) hypertrophy in adipose tissues and liver. However, the role of AG in the regulation of lipid metabolism in the intestine remains
Yadong Yu et al.
Journal of lipid research, 57(10), 1879-1888 (2016-08-20)
Human VLDLs assembled in the liver and secreted into the circulation supply energy to peripheral tissues. VLDL lipolysis yields atherogenic LDLs and VLDL remnants that strongly correlate with CVD. Although the composition of VLDL particles has been well-characterized, their 3D
Jameson T Crowley et al.
The Journal of infectious diseases, 212(11), 1841-1850 (2015-05-28)
To discover novel autoantigens associated with Lyme arthritis (LA), we identified T-cell epitopes presented in vivo by human leukocyte antigen (HLA)-DR molecules in patients' inflamed synovial tissue or joint fluid and tested each epitope for autoreactivity. Using this approach, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service