Skip to Content
Merck
All Photos(2)

Key Documents

LUXSHRTP

Millipore

mPAGE®Lux Masked Short Plate

greener alternative

Special short plate for use with MPAGE® Lux Curing Station

Synonym(s):

masked mini gel short plates

Sign Into View Organizational & Contract Pricing

Select a Size

5 EACH
$94.70
5 EACH
$94.70

$94.70


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 EACH
$94.70
5 EACH
$94.70

About This Item

UNSPSC Code:
41105312
NACRES:
NB.15

$94.70


Available to ship on07 April 2025Details


Request a Bulk Order

Product Name

mPAGE® Lux Masked Short Plates, 5pk,

material

glass plate

description

for use with mPAGE® Lux Curing Station with 10 wells or 15 wells

packaging

pkg of 5 packs (10 wells)
pkg of 5 packs (15 wells)

manufacturer/tradename

Millipore

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

W × H

10.1 cm × 7.3 cm

gel size

8.3 cm × 10.1 cm

compatibility

for use with mPAGE®

  • Lux Bis-Tris Reagent Kit
  • Lux Curing Station
  • Lux Mixing Tubes Combs
  • Mini Spacer Plates
  • Mini Gel Tank
  • Mini Wet Transfer System

greener alternative category

General description

Use mPAGE® Lux Masked Short Plates 10 wells, 5pk when casting polyacrylamide gels using mPAGE® Lux Curing Station with cassette sandwiches assembled from mPAGE® Mini Spacer Plates and mPAGE® Combs.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service