Skip to Content
Merck
All Photos(3)

Documents

IPAKGARDH2

Milli-Q

IPAK Gard® 10/15 Pretreatment Cartridge for Hard Water

For Milli-Q® IQ/IX 7010/15 water systems to protect RO membranes

Synonym(s):

IPAK Gard 10/15 packs for hard water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212

Quality Level

packaging

units of 1

manufacturer/tradename

Milli-Q®

storage condition

room temperature

cartridge H

39.5 cm (15.5 in.)

input

feed water nature tap water (within specifications of the water purification system)

compatibility

for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015

shipped in

ambient

General description

The IPAK Gard® 10/15 pretreatment cartridge for hard water protects the reverse osmosis (RO) membrane by efficiently removing water contaminants such as free chlorine, colloids, particles and minerals from hard water.
Purification technologies include activated carbon for the removal of free chlorine, polyphosphate beads to protect against scaling and precipitates, and a pleated filter for the removal of particulates and colloids. This pretreatment cartridge improves your system′s performance and extends its lifetime.

Application

  • For use with Milli-Q® IQ/IX 7010/15.
  • Optimally protects RO membranes from free chlorine, colloids, particles and minerals found in hard feed water.
  • Improves system performance and lifetime.

Features and Benefits

  • Pleated filter removes particulates and colloids, and activated carbon block removes free chlorine from hard feed water to ensure RO membrane performance over years
  • Polyphosphate beads protect against scaling and precipitates
  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with a Drain Cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal.
  • 63% of the parts suppliers are ISO 14001 environmental management system certified.

Legal Information

IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service