Skip to Content
Merck
All Photos(1)

Key Documents

IEVH07850

Millipore

Immobilon® -E PVDF Membrane

Immobilon®-E PVDF Membrane

50 sheets, 7 cm x 8.4 cm, 0.45 µm pore size, transfer membrane wets out in transfer buffer or water

Synonym(s):

Western blotting membrane, blotting membrane, mini blots, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.23
material:
PVDF membrane
plain filter
white filter
detection method:
chemiluminescent
colorimetric
radioactive
pore size:
0.45 μm pore size
compatibility:
for use with Amido black
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Transillumination
Pricing and availability is not currently available.

Product Name

Immobilon®-E PVDF Membrane, 50 sheets, 7 cm x 8.4 cm, The first PVDF transfer Membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

easily wetted with water

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter L × W

7 cm × 8.4 cm

pore size

0.45 μm pore size

compatibility

for use with Amido black
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Transillumination

detection method

chemiluminescent
colorimetric
radioactive

shipped in

ambient

General description

Filter Code: IEVH
Immobilon-E is the first PVDF membrane for Western blotting that can be wetted with with aqueous transfer buffer. This eliminates the alcohol pre-wet step required with other PVDF membranes.
Primary Binding Mechanisms: Electrostatic, hydrophobic

Application

50 sheets, 7 cm x 8.4 cm, The first PVDF transfer Membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Questions

Reviews

Active Filters

  1. New york
    • Review 1
    • Vote 1
    4 out of 5 stars.

    PVDF non alcohol activation

    effective and very easy for applications like slot blot, gel transfer

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service