Skip to Content
Merck
All Photos(1)

Documents

HX0607

Supelco

Hydrochloric Acid

34-37% OMNITRACE®

Synonym(s):

Hydrochloric acid solution, Muriatic Acid, 23 deg. Be, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
41116105

product name

Hydrochloric Acid, 34-37% OmniTrace

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8
suitable for GB 5009.253-2016
suitable for SM 5210

vapor pressure

190 hPa ( 20 °C)

form

liquid

concentration

(Nitric Acid, >= 65 - < 70)
≥30-50% in HCl

pH

<1 (20 °C in H2O)

mp

(Solidification point: -30 °C (-22 °F))

solubility

soluble (Water: Soluble)

density

ca.1.19 g/cm3 at 20 °C (68 °F)

shipped in

ambient

storage temp.

room temp

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

OmniTrace acids offer an economical alternative for sensitive, trace metal analysis and sample preparation. These acids are tested to PPB (parts per billion) levels.

Packaging

Product is packed into FEP PTFE bottles. Each bottle is packed into a single bottle shipping case.

Legal Information

OMNITRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service