Skip to Content
Merck

Skip To

HTTP03700

Polycarbonate Membrane Filter, 0.4 μm Pore Size

Isopore™, filter diam. 37 mm, hydrophilic

Sustainability features

Synonym(s):

Isopore Membrane Filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 filters
Check Cart for Availability
$269.00

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
0.4 μm pore size, 10-20 % porosity
Material:
plain filter, polycarbonate membrane, white filter
Feature:
hydrophilic
Diam.:
37 mm
Sterility:
non-sterile

$269.00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

plain filter, polycarbonate membrane, white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Isopore™, Millipore

sustainability

Sustainability features

parameter

≥50 mL/min-cm2 water flow rate at 10 psi (typical results), ≥7 L/min air flow rate at 10 psi, 140 °C max. temp., 18 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)

diam.

37 mm

filter diam.

37 mm

thickness

10 μm

gravimetric extractables

<1%

refractive index

n/D 1.6

matrix

Isopore™

pore size

0.4 μm pore size, 10-20 % porosity

bubble point

≥1.5 bar (22 psi) at 23 °C

greener alternative category

shipped in

ambient

General description

Filter Code: HTTP
Filter Type: Screen filter
The Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Absorbable organic halides (AOX)
  • Particle monitoring
  • Air monitoring
  • Microplastics analysis
  • Microplastics analysis grade water

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Possible reduction in flow rates may occur after autoclaving.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HTBP01300HTTP04700HTTP09030
description

Isopore™, filter diam. 37 mm, hydrophilic

description

Isopore™, filter diam. 13 mm, hydrophilic, brown

description

Isopore™, filter diam. 47 mm, hydrophilic, white

description

Isopore™, filter diam. 90 mm, hydrophilic

parameter

140 °C max. temp., ≥7 L/min air flow rate at 10 psi, ≥50 mL/min-cm2 water flow rate at 10 psi (typical results), 18 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)

parameter

140 °C max. temp., ~49 mL/min-cm2 water flow rate at 10 psi (typical results), 18 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)

parameter

140 °C max. temp., ≥7 L/min air flow rate at 10 psi, ≥50 mL/min-cm2 water flow rate at 10 psi (typical results), 18 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)

parameter

≥50 mL/min-cm2 water flow rate at 10 psi (typical results), 18 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), ≥7 L/min air flow rate at 10 psi, 140 °C max. temp.

pore size

0.4 μm pore size, 10-20 % porosity

pore size

5-20 % porosity, 0.4 μm pore size

pore size

0.4 μm pore size, 10-20 % porosity

pore size

0.4 μm pore size, 10-20 % porosity

diam.

37 mm

diam.

13 mm

diam.

47 mm

diam.

90 mm

manufacturer/tradename

Millipore, Isopore™

manufacturer/tradename

Millipore, Isopore™

manufacturer/tradename

Isopore™, Millipore

manufacturer/tradename

Millipore, Isopore™

material

polycarbonate membrane, white filter, plain filter

material

brown filter, polycarbonate membrane, plain filter

material

polycarbonate membrane, plain filter, white filter

material

polycarbonate membrane, plain filter, white filter

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters