Skip to Content
Merck
All Photos(1)

Key Documents

HTS209RTA

Sigma-Aldrich

Ready-to-Assay LPA 5/GPR92 Lysophospholipid Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
Product HTS209RTA is not currently sold in your country. Contact Technical Service

Product Name

Ready-to-Assay LPA 5/GPR92 Lysophospholipid Receptor Frozen Cells,

biological source

human

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Lysophosphatidic acid (LPA) is a lysophospholipid produced by activated platelets that inhibits adenylate cyclase and stimulates DNA synthesis, changes in cell morphology, and increases in intracellular calcium in a variety of cultured mammalian cells. A growing family of GPCRs mediates the biological effects of LPA. The first LPA receptors described, LPA1-3, share relatively high sequence similarity and are related to the sphingosine 1-phosphate receptors S1P1-5 (Contos et al., 2000). Two more recently characterized LPA receptors, LPA4 and LPA5, are related to each other but are more distantly related to LPA1-3. LPA5, originally known as GPR92, mediates LPA-induced cytoskeletal changes, intracellular calcium flux and increased cAMP by coupling to G12/13, Gq, and Gs. (Lee et al., 2006). In addition to binding LPA, LPA5 is also activated by several other lipid-derived molecules, the most potent of which is farnesyl pyrophosphate (Oh et al., 2008). GPR92 is expressed in several tissues, most prominently in the CD8+ intraepithelial lymphocytes of the gastrointestinal tract (Kotarsky et al., 2006). Millipore′s cloned human LPA5-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant LPA5 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at LPA5.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human LPA5 / GPR92 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: LPA5 / GPR92
  • Target Sub-Family: Lysophospholipid

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service