Skip to Content
Merck
All Photos(1)

Key Documents

HTS172RTA

Sigma-Aldrich

Ready-to-Assay CALCRL Calcitonin Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
Product HTS172RTA is not currently sold in your country. Contact Technical Service

Product Name

Ready-to-Assay CALCRL Calcitonin Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

The calcitonin family of peptides, which includes calcitonin, two calcitonin-related gene peptides (CGRPs), adrenomedullin, and amylin, share a disulfide-bonded loop, an amphipathic a-helix, and an amidated C-terminus. The peptides influence a variety of physiological effects, including vascular tone, food intake and bone metabolism. The biological effects of the peptides are mediated by two class B GPCRs, CT receptor (calcitonin receptor) and CL receptor (calcitonin receptor-like receptor) (Poyner et al., 2002). The pharmacology of the CT and CL receptors is strongly influenced by their association with a family of three receptor activity modifying proteins (RAMPs), which are 14-17 kD single pass transmembrane proteins. CGRP (calcitonin gene-related peptide) induces vasodilation, and its activity is primarily mediated by CGRP1, composed of CL in complex with RAMP1 (Hasbak et al., 2003). As a consequence of its role as a vasodilator, CGRP acting via CGRP1 appears to play an important role in hypertension (Deng and Li, 2005). Millipore′s cloned human CALCRL -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant CALCRL expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at CALCRL.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human CGRP1 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: B
  • Protein Target: CGRP1
  • Target Sub-Family: Calcitonin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service