Skip to Content
Merck
All Photos(2)

Key Documents

HTS121M

Sigma-Aldrich

ChemiSCREEN Human Motilin Receptor Membrane Preparation

Human motilin receptor / MTLR / GPR38 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
$1,350.00

$1,350.00


Estimated to ship on25 May 2025



Select a Size

Change View
200 UNITS
$1,350.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

$1,350.00


Estimated to ship on25 May 2025


biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Human Motilin Receptor
Motilin is a 22 amino acid peptide that potently stimulates gastrointestinal contractility. The biological effects of motilin are mediated by a Gq-coupled seven transmembrane protein, currently termed motilin receptor (MR), that shares significant sequence similarity with the ghrelin receptor (Feighner et al., 1999). The motilin receptor is also activated by the antibiotic erythromycin; this interaction appears to mediate some of the gastrointestinal side effects of erythromycin. Although motilides (non-antibiotic derivatives of erythromycin) such as ABT-229 have been investigated for treatment of diabetic gastroporesis, the effectiveness has been limited by tachyphylaxis (decreased response to ligand) resulting from receptor downregulation (Thielemans et al., 2005). Agonists of the motilin receptor with reduced densensitization activity remain a potential treatment for disorders of gastric motility. Chemicon′s cloned human Motilin Receptor-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant Motilin Receptor expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between the Motilin Recptor and its ligands. Chemicon′s motilin membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of motilin receptor interactions with motilin. The membrane preparations exhibit a Kd of 0.43nM for [125I]-Motilin. With 5.0 µg/well Motilin Membrane Prep and 0.5 nM [125I]-Motilin, a greater than 30-fold signal-to-background ratio was obtained.

Application

Radioligand binding assay, and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: motilin receptor / MTLR / GPR38
Target Sub-Family: Motilin

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of Motilin Receptor membrane prep:
5 µg/well10 µg/well
Signal:Background 75 65
Specific Binding (cpm) 4634854457


SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax: 5.6 pmol/mg
Kd: 0.43 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I] Motilin (Perkin Elmer # NEX378)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 30-fold signal:background with 125I-labeled motilin at 0.5 nM.

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein were adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desensitization of the human motilin receptor by motilides.
Thielemans, Leen, et al.
Journal of Pharmacology and Experimental Therapeutics, 313, 1397-1405 (2005)
S D Feighner et al.
Science (New York, N.Y.), 284(5423), 2184-2188 (1999-06-26)
Motilin is a 22-amino acid peptide hormone expressed throughout the gastrointestinal (GI) tract of humans and other species. It affects gastric motility by stimulating interdigestive antrum and duodenal contractions. A heterotrimeric guanosine triphosphate-binding protein (G protein)-coupled receptor for motilin was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service