HTS103M
ChemiSCREEN Human D3 Dopamine Receptor Membrane Preparation
Human D3 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
biological source
human
Quality Level
recombinant
expressed in Chem-1 cells
manufacturer/tradename
ChemiScreen
Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
Dopamine is a catecholamine neurotransmitter that functions in the CNS to control locomotor, cognitive, emotional and neurendocrine processes, and in the periphery to modulate cardiovascular, renal and gastrointestinal processes. The biological activities of dopamine are mediated by a family of five GPCRs. The D1 and D5 subtypes couple to Gs to increase intracellular cAMP, whereas the D2, D3 and D4 subtypes couple to Gi to reduce cAMP (Missale et al., 1998). The dopamine D3 receptor is expressed in phylogenetically older regions of the brain, suggesting that it plays a role in cognitive and emotional functions. It has been the target of potential drugs for psychotic disorders and Parkinson′s disease. Chemicon′s D3 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of D3 interactions and its ligands. The membrane preparations exhibit a Kd of 0.1 nM for [125I]-Iodospiperone. With 5.0ug/well D3 membrane prep and 0.1 nM [125I]-Iodospiperone, a greater than 4-fold signal-to-background ratio was obtained.
Human D3 Receptor
Application
Human D3 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Biochem/physiol Actions
Protein Target: D3
Quality
Table 1. Signal:background and specific binding values obtained in a competition binding assay with varying amounts of D3 Receptor membrane prep.
SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax: 0.32 pmol/mg
Kd: 0.1 nM
10 µg/well | 5 µg/well | |
---|---|---|
Signal:Background | 4.88 | 5.93 |
Specific Binding (cpm) | 1853.7 | 1861.3 |
SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax: 0.32 pmol/mg
Kd: 0.1 nM
Specifications
Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM Tris, pH 7.4. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Tris, pH 7.4, 120mM NaCl, filtered and stored at 4°C
Radioligand: [125I] -Iodospiperone(Perkin Elmer # NEX284)
Wash Buffer: 50 mM Tris, pH 7.4, filtered and stored at 4°C.
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM Tris, pH 7.4. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Tris, pH 7.4, 120mM NaCl, filtered and stored at 4°C
Radioligand: [125I] -Iodospiperone(Perkin Elmer # NEX284)
Wash Buffer: 50 mM Tris, pH 7.4, filtered and stored at 4°C.
Physical form
One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 4-fold signal:background with 125I-labeled Iodospiperone at 0.1 nM.
Liquid in packaging buffer: 50 mM Tris, pH 7.4, 10% glycerol with no preservatives.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80oC.
Liquid in packaging buffer: 50 mM Tris, pH 7.4, 10% glycerol with no preservatives.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80oC.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service