Skip to Content
Merck
All Photos(1)

Key Documents

HTS043RTA

Sigma-Aldrich

Ready-to-Assay VPAC1 VIP and PACAP Receptor Frozen Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
Product HTS043RTA is not currently sold in your country. Contact Technical Service

description

Cell line type: GPCR Cell Lines
GPCR Class B
protein target VPAC1 / VIP1

Quality Level

technique(s)

calcium flux assay: suitable

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Vasoactive intestinal peptide (VIP), a 28 amino acid peptide originally isolated by its vasodilation activity, binds to two class B GPCRs, VPAC1 and VPAC2, to exert its functions in the CNS, vasculature, immune system and adrenal medulla (Harmar et al., 1998). In the immune system, VIP is synthesized by mast cells and lymphocytes, and appears to inhibit inflammation and to shift the immune response toward a Th2 pathway (Delgado et al., 2004). In the heart, VIP is expressed by nerve fibers, where it modulates heart rate, and coronary blood flow (Henning and Sawmiller, 2001). Millipore′s cloned human VPAC1-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant VPAC1 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at VPAC1.

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service