Skip to Content
Merck
All Photos(1)

Key Documents

GFCP083000

Millipore

Glass fiber Conjugate Pad 8mm x 300mm strips 100 pk

Sign Into View Organizational & Contract Pricing

Select a Size

100 STRIPS
$53.70

$53.70


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 STRIPS
$53.70

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

$53.70


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service