Skip to Content
Merck
All Photos(1)

Key Documents

GF35L

Sigma-Aldrich

Anti-BDNF Mouse mAb (35928.11)

lyophilized, clone 35928.11, Calbiochem®

Synonym(s):

Anti-Brain Derived Neurotrophic Factor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$608.00

$608.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μG
$608.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$608.00


Estimated to ship on30 May 2025


biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

35928.11, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

General description

Anti-BDNF, mouse monoclonal, clone 35928.11, recognizes the ~20 kDa BDNF protein in brain extracts. It is validated for ELISA, Western blotting, immunohistochemistry, and neutralization studies.
Immunoaffinty purified mouse monoclonal antibody. Recognizes the ~20 kDa BDNF protein.
Recognizes the ~20 kDa BDNF protein in brain extracts. Clone 35928.11 was selected based on its ability to neutralize the biological activity of human recombinant BDNF.

Immunogen

recombinant protein consisting of amino acids 129-247 of human BDNF

Application

ELISA (0.5-1 µg/ml)

Immunoblotting (1-2 µg/ml)

Immunohistochemistry (5-15 µg/ml)

Neutralization Studies (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from PBS, 5% trehalose.

Reconstitution

We recommend resuspending the lyophilized antibody with sterile PBS, pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml. Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Riccio, A., et al. 1997. Science277 1097.
Sendtner, M., et al. 1996. Neurochem. Res.21, 831.
Barbacid, M. 1995. Ann. N.Y. Acad. Sci.766, 442.
Gotz, R., et al. 1994. Nature372, 266.
Snider, W.D. 1994. Cell77, 627.
Snider, W.D., et al. 1992. J. Neurobiol.23, 1231.
The full-length sequence for BDNF can be found under accession number NP_001700. This antibody has been selected for its ability to neutralize the biological activity of human recombinant BDNF. The exact concentration of antibody required to neutralize human recombinant BDNF activity is dependent on the cytokine concentration, cell type, growth conditions, and the type of activity studied. Suggested neutralization concentration required to yield one-half maximal inhibition of BDNF activity is approximately 5-15 µg/ml in the presence of 2.5 ng/ml of human recombinant BDNF, in a neuron survival assay using embryonic chick dorsal root ganglia neurons. The detection limit for human recombinant BDNF is ~300 ng/lane under non-reducing and reducing conditions. The detection for human recombinant BDNF is ~12.5 ng/ml. Based on immunoblotting and ELISA, this antibody exhibits less than 2% cross-reactivity with human recombinant β-NGF, human recombinant NT-3, and human recombinant NT-4. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Markham et al.
The European journal of neuroscience, 20(5), 1189-1196 (2004-09-03)
Brain-derived neurotrophic factor (BDNF) governs both the selective survival of neurons during development and the experience-based regulation of synaptic strength throughout life. BDNF produced a concentration-dependent increase in the respiratory control index (RCI, a measure of the efficiency of respiratory
Natalia A Stefanova et al.
Aging, 8(11), 2713-2733 (2016-10-18)
Mitochondrial aberrations are observed in human Alzheimer's disease (AD) and in medical conditions that increase the risk of this disorder, suggesting that mitochondrial dysfunction may contribute to pathophysiology of AD. Here, using OXYS rats that simulate key characteristics of sporadic
Tinmarla F Oo et al.
Molecular and cellular neurosciences, 41(4), 440-447 (2009-05-05)
Brain-derived neurotrophic factor (BDNF) was the first purified molecule identified to directly support the development of mesencephalic dopamine neurons. However, its physiologic role has remained unknown. Based on patterns of expression, it is unlikely to serve as a target-derived neurotrophic
Hugo Talbot et al.
Scientific reports, 10(1), 12572-12572 (2020-07-30)
Evading apoptosis and sustained survival signaling pathways are two central hallmarks of B-cell chronic lymphocytic leukemia (B-CLL) cells. In this regard, nurse-like cells (NLC), the monocyte-derived type 2 macrophages, deliver stimulatory signals via B-cell activating factor (BAFF), a proliferation-inducing ligand
Milena Penkowa et al.
Journal of neuroscience research, 79(4), 522-534 (2004-12-23)
We examined metallothionein (MT)-induced neuroprotection during kainic acid (KA)-induced excitotoxicity by studying transgenic mice with MT-I overexpression (TgMT mice). KA induces epileptic seizures and hippocampal excitotoxicity, followed by inflammation and delayed brain damage. We show for the first time that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service