Skip to Content
Merck
All Photos(1)

Documents

GF023

Sigma-Aldrich

Tumor Necrosis Factor-α Protein, Recombinant human

Tumor Necrosis Factor-alpha (TNF-alpha) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells & certain other target cells.

Synonym(s):

TNF-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

biological source

human

Quality Level

Assay

>97% (SDS-PAGE)

specific activity

≥2 x 107 U/mg

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

input

sample type hematopoietic stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... TNF(7124)

General description

Product Source: E.coli
Tumor Necrosis Factor-alpha (TNF-alpha) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. The mature form of human TNF-alpha has 157 amino acid residues. GF023 is a recombinant polypeptide containing the mature form of human TNF-a with a polyhistidine tag at its amino terminal.

Application

For most in vitro applications, TNF-alpha exerts its biological activity in the concentration range of 0.05 to 20.0 ng/mL. Responding cells are (partial list): Tumor cells, certain other cells.

Physical form

Lyophilized from 3mM Tris, pH 8.0.

Storage and Stability

The lyophilized powder, though stable at room temperature, is best stored desiccated at -20°C. Reconstitute with sterile distilled water to a concentration of 0.1-1 mg/mL. Note: Allow the reconstituted vial to sit at room temperature for 2 hours before use. This solution can be diluted into other buffered solutions and stored at 4°C for one week or -20°C for future use. Reconstituted TNF-alpha should be stored in undiluted aliquots at -20°C for up to six months.

Analysis Note

Specific Activity: Chemicon′s human TNF-alpha, is fully biologically active when compared to standards.The ED50 as determined by the cytolysis of mouse L929 cells in the presence of Actinomycin-D is ≤ 0.05 ng/mL

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Angélica Contreras et al.
The Biochemical journal, 477(17), 3299-3311 (2020-09-18)
TNFα is a pro-inflammatory cytokine that is a therapeutic target for inflammatory autoimmune disorders. Thus, TNFα antagonists are successfully used for the treatment of these disorders. Here, new association patterns of rhTNFα and its antagonists Adalimumab and Etanercept are disclosed.
Sahar Rafat et al.
Biology, 11(11) (2022-11-12)
Cancer is the utmost common disease-causing death worldwide, characterized by uncontrollable cell division with the potential of metastasis. Overexpression of the Inhibitors of Apoptosis proteins (IAPs) and autophagy correlates with tumorigenesis, therapeutic resistance, and reoccurrence after anticancer therapies. This study
María A Contreras et al.
Proteins, 89(11), 1557-1564 (2021-07-13)
The neutralization of tumor necrosis factor alpha (TNFα) with biopharmaceuticals is a successful therapy for inflammatory diseases. Currently, one of the main TNFα-antagonists is Etanercept, a dimeric TNF-R2 ectodomain. Considering that TNFα and its receptors are homotrimers, we proposed that
Hangeun Kim et al.
Methods in molecular biology (Clifton, N.J.), 1155, 125-132 (2014-05-03)
Hepatitis C virus (HCV) often causes chronic infection in humans, although the mechanisms for viral chronicity are not clearly understood. Tumor necrosis factor-α (TNF-α)-mediated apoptosis is a key element in a host organism's defense inhibiting viral spread and persistence. HCV
Rojo A Ratsimandresy et al.
Nature communications, 8, 15556-15556 (2017-06-06)
Inflammasomes are protein platforms linking recognition of microbe, pathogen-associated and damage-associated molecular patterns by cytosolic sensory proteins to caspase-1 activation. Caspase-1 promotes pyroptotic cell death and the maturation and secretion of interleukin (IL)-1β and IL-18, which trigger inflammatory responses to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service