Skip to Content
Merck
All Photos(1)

Key Documents

FX0437

Supelco

Acetonitrile solution

suitable for HPLC, contains 0.1% (v/v) formic acid

Synonym(s):

Acetonitrile solution, ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide, acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
4124158
MDL number:
UNSPSC Code:
12352117
EC Index Number:
200-835-2

Product Name

Acetonitrile/Formic Acid 0.1% Mobile Phase,

form

liquid

Quality Level

concentration

0.095-0.105 % (v/v) CH3COOH

evapn. residue

≤5 ppm

color

APHA: ≤10

transition temp

flash point 5.9 °C

density

0.782 g/mL at 25 °C

format

mixture

shipped in

ambient

storage temp.

room temp

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

FX0437P-1 is packaged in poly coated glass bottles

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Assay (HCOOH)0.095-0.105% (v/v)Color (APHA)10 maxDensity at 25°CDetermineg/mLEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidGradient at 280 nm0.005AU maxUV Abs. at 230 nm1.00AU maxUV Abs. at 240 nm0.50AU maxUV Abs. at 250 nm0.05AU maxUV Abs. at 280 nm and higher0.05AU max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.6 °F

Flash Point(C)

5.9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service