Skip to Content
Merck
All Photos(1)

Key Documents

FRZ0020L002P5

Millipore

Mobius® Gold 2D Freeze Assembly w/Helium IT

20L PureFlex film with Lynx® S2S connectors

Synonym(s):

Mobius®, Gold w/Helium IT, 20L PF FRZ TRN Assy w/ [LM="Lynx"] S2S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
Pricing and availability is not currently available. Contact Technical Service

material

C-Flex® tubing (374)
PureFlex device (film)
connections (Lynx S2S male and female connector
Lynx S2S male and female connector)

Quality Level

Agency

certified by the (Assembly performed under ISO Clean Room Class 7 conditions in a facility)
certified by the ISO (Bags manufactured under ISO Clean Room Class 8 conditions in a facility)

sterility

sterile; γ-irradiated (level of 25-40 kGY)

product line

Mobius® Freeze

packaging

package of 5 ×

parameter

-80-60 °C temp. range (112-140 °F)
20 L process volume

Looking for similar products? Visit Product Comparison Guide

General description

The Mobius® 2D freeze single-use assembly offers a robust freeze and thaw solution that allows safe storage and transportation of your valuable product at a very low temperature while maintaining the integrity of the final product. Combined with the RoSS® (Robust Storage and Shipping) shell, this solution enables easy and safe bulk drug substance and product transfer between multiple sites providing flexibility and security to your processes. Available in 1L, 5L, 10L, and 20L sizes, this standard 2D Freeze assembly is made with our PureFlex film and is tested for use at temperatures down to - 80 °C during, freezing, storage, transportation, and thawing.

Application

Freeze/Thaw Applications
Protect and Freeze Your Bulk Drug Substance

Packaging

Five (5) assembies per package with each assembly containing five (5) 20L bags

Components

PureFlex filmC-Flex 374 tubingLynx S2S male and female connector NovaSeal pinch pipe

Other Notes

• Mobius® 2D Freeze Assemblies are intended to be used solely with Single Use Support RoSS® shells.• User shall be solely responsible and liable for validation and use of the Mobius® bag assemblies beyond the range of our specifications.• Carefully handle the Mobius® assembly at all times.• Do not handle the Mobius assembly until completely thawed.

Legal Information

C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
LYNX is a registered trademark of Merck KGaA, Darmstadt, Germany
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany
Ross is a registered trademark of Thermo Electron Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service