Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB404PE

Sigma-Aldrich

Milli-Mark® Anti-Tie2/TEK-PE Antibody, clone Ab33

clone Ab33, Milli-Mark®, from mouse

Synonym(s):

Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, hTIE2, p140 TEK, CD202b

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$381.00

$381.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$381.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

Ab33, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TEK(7010)

Related Categories

General description

TIE2 (tyrosine kinase with Ig and EGF homology domains 2) is expressed almost exclusively in endothelial cells in mice, rats and humans. This receptor possesses a unique extracellular domain containing two immunoglobulin-like loops separated by three epidermal growth factor-like repeats that are connected to three fibronectin type III-like repeats. The ligand for the receptor is Angiopoietin 1. Defects in TIE2 are associated with inherited venous malformations; the TIE2 signaling pathway appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis.

Specificity

Antibody recognizes Human Tie2/TEK.

Immunogen

Fusion protein corresponding to residues 1-745 of human Tie2/TEK with a C-terminal His6-tag purified from infected SF-9 cells.

Application

Milli-Mark Anti-Tie2/TEK-PE Antibody, clone Ab33 is an antibody against Tie2/TEK for use in FC.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Evaluated by flow cytometry using HUVEC cells.

Target description

126 kDa Calculated

Physical form

Protein G Purified
Purified mouse monoclonal IgG1k conjugated to PE in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
HUVEC cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason O Moore et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(17), 4382-4387 (2017-04-12)
Tie1 and Tie2, members of the tyrosine kinase family with immunoglobulin and EGF homology domains, are receptor tyrosine kinases found primarily in endothelial cells with key roles in development and maintenance of the vasculature and in angiogenesis. They are attractive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service