Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB390F

Sigma-Aldrich

Milli-Mark® Anti-pan Histone-FITC Antibody, clone H11-4

clone H11-4, Milli-Mark®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$386.00

$386.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
$386.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$386.00


Estimated to ship on08 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

H11-4, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H3C1(8350)

Related Categories

General description

There are 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. The five major classes of histones are H1/H5, H2A, H2B, H3, and H4. Histones H2A, H2B, H3 and H4 are known as the core histones, while histones H1 and H5 are known as the linker histones. Histones feature a main globular domain and a long N terminal tail which are involved with the structure of the nucleosomes of the ′beads on a string′ structure. They are the chief protein components of chromatin, acting as spools around which DNA winds, and play a role in gene regulation.

Specificity

Due to the highly conserved nature of histones, this antibody is predicted to have wide species reactivity.

Application

This Milli-Mark Anti-pan Histone-FITC Antibody, clone H11-4 is validated for use in FC for the detection of pan Histone.

Quality

Evaluated by flow cytometry using HeLa cells.

Target description

Antibody recognizes histones H1, H2A, H2B, H3, and H4. Calculated molecular weight for these histones range from 14 to 21 kDa

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service