Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB288A6

Sigma-Aldrich

Milli-Mark® Anti-Ubiquitin Antibody, Lys63-Specific-Alexa Fluor647, clone Apu3

clone Apu3, Milli-Mark®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$583.00

$583.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$583.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

$583.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Apu3, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBB(7314)

General description

Polyubiquitin chains linked through the Lys48 residue of ubiquitin are most commonly associated with proteins targeted for proteosomal degradation. In contrast, polyubiquitin chains linked through the Lys63 residue of ubiquitin (Ub) are associated with nonproteolytic functions such as signal transduction. Lys63-linked polyUb chains connect components of NFκB signaling in a highly regulated manner, and genetic evidence indicates the involvement of Lys63-linked Ub chains in stress response and DNA repair. Research in yeast has suggested Lys63-linked Ub chains stimulate endocytosis.

Specificity

Recognizes ubiquitin linked at Lys63. Does not recognize ubiquitin linked at Lys48 or free ubiquitin.

Immunogen

Immunogen used was di-ubiquitin linked at Lys63.

Application

This Milli-Mark Anti-Ubiquitin Antibody, Lys63-Specific-Alexa Fluor647, clone Apu3 is validated for use in FC for the detection of Ubiquitin.

Quality

Evaluated by flow cytometry using HeLa cells

Target description

Molecular weight varies.

Physical form

Purified recombinant rabbit monoclonal IgG1 conjugated to Alexa Fluor 647 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service