Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB216AP

Sigma-Aldrich

Milli-Mark® Anti-CD138-APC Antibody, clone MI15

clone MI15, Milli-Mark®, from mouse

Synonym(s):

Syndecan-1, SYND1

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$507.00

$507.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
$507.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$507.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MI15, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SDC1(6382)

General description

CD138 is a transmembrane proteoglycan, bearing heparin sulphates and sometimes chondriotin sulfates. It consists of a 30.5 kDa core protein with 5 putative glycosaminoglycans attachment sites. CD138 is a member of the syndecan family. The reported Mr of CD138 ranges from 85,000 to 92,000, presumably depending on the glycosaminoglycan pattern of the molecule (Wijdenes, 1997). Within the hematopoietic system, CD138 is mainly confined to late stages of B-cell differentiation. It is typically expressed at high cellular density in plasma cells (Gattei, 1999). In mature tissues, CD138 is expressed by simple and stratified epithelia, fibroblasts, stratified keratinocytes and endothelial cells (Wijdenes, 1997). Anti-CD138, MI15, was included in the Sixth International Workshop and Conference on Human Leucocytes Differentiation Antigens, and studies by a number of different laboratories confirmed its reactivity with CD138 (Wijdenes, 1997).

Specificity

Antibody recognizes Human CD138 antigen.

Immunogen

A mixture of U266 and XG-1 human myeloma cell lines

Application

Detect CD138-APC using this Milli-Mark Anti-CD138-APC Antibody, clone MI15 validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using U266 cells

Target description

85-92 kDa (Wijdenes, 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to APC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
U266 cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service