Skip to Content
Merck
All Photos(2)

Key Documents

EZHAGG504

Millipore

EZ-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, green filter, gridded filter, General microbiological analysis

Synonym(s):

Membrane filters, Semi-permeable membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available. Contact Technical Service

material

green filter
gridded filter
mixed cellulose esters (MCE)

Quality Level

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic
individually wrapped

manufacturer/tradename

EZ-Pak®

filter diam.

50 mm

matrix

MF-Millipore

pore size

0.45 μm pore size

input

sample type wine
sample type soft drink
sample type cosmetics
sample(s) (environmental)
sample type beer
sample type food(s)
sample type sport drink
sample type beverage(s)
sample type cider
sample type bottled water
sample type pharmaceutical(s)

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

shipped in

ambient

Related Categories

General description

EZ-Pak Filters are made of Mixed cellulose esters (MCE) with 0.45 μm pore size, green gridded and 50mm diameter. These filters possess high recoveries and are more appropriate for general microbiological testing. EZ-Pak membranes and dispenser revolutionize filter handling for Membrane Filtration (MF) method testing by combining the benefits of ready-to-use, sterile, individually packed membranes with unique handling ease. Green membranes are ideal for spotting small, clear, translucent colonies

Application

The EZ-Pak system is suitable for testing drinking, bottled and mineral water according to international standards. These filters find application in Cosmetics, Environmental and Food and Beverage industry. Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method.

Legal Information

EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service