Skip to Content
Merck
All Photos(2)

Key Documents

EZAAWG474

Millipore

EZ-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, gridded white filter, suitable for bioburden testing, input: beverage(s)

Synonym(s):

Membrane filters, Semi-permeable membranes

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 150 FILTERS
$787.00

$787.00


Check Cart for Availability

Request more information

Select a Size

Change View
4 X 150 FILTERS
$787.00

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

$787.00


Check Cart for Availability

Request more information

material

gridded filter
gridded white filter
mixed cellulose esters (MCE)

Agency

ISO 7704

description

Yeast or mold

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic
individually wrapped

manufacturer/tradename

EZ-Pak®

filter diam.

47 mm

matrix

MF-Millipore

pore size

0.8 μm pore size

input

sample type beverage(s)
sample type cosmetics
sample type beer
sample(s) (environmental)
sample type pharmaceutical(s)
sample type sport drink
sample type soft drink
sample type wine
sample type bottled water
sample type cider
sample type food(s)

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Related Categories

General description

EZ-Pak Filters are made of Mixed cellulose esters (MCE) with 0.8 μm pore size, white, gridded and 47mm diameter. EZ-Pak membranes and dispenser revolutionize filter handling for Membrane Filtration (MF) method testing by combining the benefits of ready-to-use, sterile, individually-packed membranes with unique handling ease. These filters are widely used to recover yeast and mold from samples

Application

Membrane filtration method for microbial analysis of beverages such as water, wine, beer, sport drinks.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

A touch-free membrane filter dispenser can streamline your membrane filtration testing workflow. Use with EZ-Pak® filter membranes for high-volume sampling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service