EPA100
EPA Peroxidase Protection Buffer
500 mL EPA Peroxidase Protection Buffer for ELISA.
Select a Size
$696.00
Estimated to ship on16 April 2025
Select a Size
About This Item
$696.00
Estimated to ship on16 April 2025
Recommended Products
form
liquid
packaging
pkg of 500 mL
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
shipped in
wet ice
General description
Application
Chemicon HRP conjugated antibodies may be diluted up to 1:16,000 in Peroxidase Protective Buffer. Diluted conjugates are stable for at least 3 months when stored at 2 - 8oC. The buffer is suitable for direct use in ELISA. It is recommended that the HRP conjugated antibody is diluted with Peroxidase Protective Buffer and allowed to stand for 24 hours at 2-8°C to attain constant stability before use.
Peroxidase Protective Buffer is compatible with blocking agents such as skim milk powder, Tween 20 and normal serum. However HRP conjugates should not be stored in the presence of these blocking agents. They should be added just prior to use in the assay.
Physical form
Storage and Stability
PRECAUTIONS
HRP conjugates should be diluted and stored under clean conditions to reduce the level of microbial challenge.
Sodium azide must NOT be added to Peroxidase Protective Buffer.
Avoid contamination of HRP conjugate solutions with heavy metal ions, aromatic chlorocarbons or hypochlorous acids. Avoid containers made of soda glass or polystyrene. Storage of diluted conjugates in borosilicate glass is recommended.
Efficient washing steps should follow HRP conjugate incubations. In ELISA, at least four thorough wash cycles are recommended.
Legal Information
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service