Skip to Content
Merck
All Photos(1)

Key Documents

ECM535

Sigma-Aldrich

α/β Integrin-mediated Cell Adhesion Array Combo Kit, fluorimetric

The Alpha/Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface.

Synonym(s):

Alpha/beta integrin cell adhesion kit

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$1,780.00

$1,780.00


Estimated to ship on29 May 2025



Select a Size

Change View
2 EA
$1,780.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.32

$1,780.00


Estimated to ship on29 May 2025


Quality Level

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

NCBI accession no.

detection method

fluorometric

shipped in

wet ice

Gene Information

human ... ITGA1(3672)

Application

Research Category
Cell Structure
The Alpha/Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface.
The CHEMICON® Alpha/Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface. This assay can substitute for FACS analysis (6) and is useful for screening human adherent cells for cell surface integrins or for monitoring in vitro cell differentiation or genetic modification of cells.

For Research Use Only. Not for use in diagnostic procedures.

Packaging

96 wells each

Components

Alpha Integrin Array Plate: (PN: 90599) One 96-well plate with 12 strips. Each strip contains one well each of the mouse anti-alpha integrin monoclonal antibodies: (alpha1, alpha2, alpha3, alpha4, alpha5, alphaV, alphavbeta3), and one goat anti-mouse negative well. See the Alpha Integrin plate layout on the insert data sheet.

Beta Integrin Array Plate: (PN: 90600) One 96-well plate with 12 strips. Each strip contains one well each of the mouse anti-beta integrin monoclonal antibodies: (beta1, beta2, beta3, beta4, beta6, alphaVbeta5, and alpha5beta1), and one goat anti-mouse negative well. See the Beta Integrin plate layout on the insert data sheet.

4X Cell Lysis Buffer: (Part No. 90130) One bottle - 16 mL

CyQuant GR Dye: (Part No. 90132) Two vials - 75 μL each

Assay Buffer: (Part No. 90601) Two bottles - 100 mL each

Storage and Stability

The Alpha and Beta Integrin Array Plates can be stored at 2° to 8°C in the foil pouch up to their expiration date. Unused strips may be placed back in the pouch for storage. Ensure that the desiccant remains in the pouch, and that the pouch is securely closed. Keep the remaining kit components at 2° to 8°C.

Precautions

No data is available on the biological toxicity of CyQuant GR dye. This reagent binds to nucleic acids, and as such, should be treated as a potential mutagen which may cause cancer and heritable genetic damage. Handle with caution. The DMSO stock solution should be handled with special caution as DMSO can facilitate the entry of organic molecules into tissues.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
GR is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P G Dedes et al.
Biochimica et biophysica acta, 1820(12), 1926-1939 (2012-08-14)
The extracellular matrix (ECM) components play key roles in the multistep process of cancer growth and progression. Preclinical and clinical data show that bisphosphonates (BPs) may exert direct or indirect antitumoral effects. Despite proven efficiency in cancer treatment, the mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service