Skip to Content
Merck
All Photos(1)

Key Documents

CX2290

Supelco

Cyclohexane

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Hexahydrobenzene, Cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-806-2
Assay:
≥99% (GC)
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)
Pricing and availability is not currently available.

Agency

meets USP/NF

Quality Level

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

impurities

≤0.02% Water (H2O)

evapn. residue

≤0.002%

color

clearAPHA: ≤10

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

flash point -18 °C

solubility

0.05 g/L

density

0.779 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Appearance: Clear
Assay (GC): 99.0% min
Color (APHA): 10 max
Evaporation residue: 0.002% max
Form: Liquid
Infrared Spectrum: Conforms to standard
Substances darkened by sulfuric acid: To pass test
Water (H2O): 0.02% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service