CW0671SB1
Bevigard™-M Cartridge Filter
pore size 0.5 μm, cartridge nominal length 10 in. (25 cm), Code 7 (2-226; O-ring and spear)
Synonym(s):
Bevigard™-M Cartridge Filter 10 in. Code 7 0.5 μm
About This Item
material
mixed cellulose esters (MCE)
polyester support
polypropylene
silicone seal
reg. compliance
meets EU framework regulation [1935/2004/EC] (all component materials; regarding materials and articles intended to contact food)
meets FDA Indirect Food Additive requirements cited in 21 CFR 177-182 (all component materials)
manufacturer/tradename
Bevigard™
parameter
3.5 bar max. differential pressure (50 psid) at 25 °C
cartridge nominal length
10 in. (25 cm)
diam.
7.1 cm (2.8 in.)
filtration area
0.8 m2
impurities
≤0.5 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)
gravimetric extractables
≤50 mg/cartridge
matrix
Bevigard™-M
pore size
0.5 μm nominal pore size
0.5 μm pore size
input
sample type bottled water
sample type beverage(s)
sample type beer
sample type wine
sample type liquid
sample type food(s)
cartridge code
Code 7 (2-226; O-ring and spear)
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
Packaging
Other Notes
- Organism Retention: Microorganism
- Mode of Action: Filtration (size exclusion)
- Application: BioProcessing
- Intended Use: Reduction or removal of microorganism/bioburden
- Instructions for Use: Please refer to installation and wetting sections of OptiScale®-25 user guide and introduction section of Mixed celulose esters Tech Brief
- Storage Statement: Please refer Mixed cellulose esters Tech Brief
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Specifications
Preparation Note
30 min with hot water @ 80 °C
10 autoclave cycles or SIP @ 121 °C for 30 min
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service