Skip to Content
Merck
All Photos(1)

Key Documents

CSA001-2

Sigma-Aldrich

Rho Kinase (ROCK) Activity Assay (-20°C

Sign Into View Organizational & Contract Pricing

Select a Size

CSA001-2
$542.00

$542.00


Estimated to ship on13 April 2025



Select a Size

Change View
CSA001-2
$542.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

$542.00


Estimated to ship on13 April 2025


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service