Skip to Content
Merck
All Photos(4)

Key Documents

CFSP223000

Millipore

Cellulose Absorbent Sample Pads

thickness 0.83 mm, filter L × W 20 cm × 30 cm, pack of 100 sheets

Synonym(s):

Absorbent pad, C083 Cellulose Fiber Sample Pad Sheets, Cellulose pad, Sample pad

Sign Into View Organizational & Contract Pricing

Select a Size

100 SHEETS
$435.00

$435.00


Available to ship on14 April 2025Details


Request more information

Select a Size

Change View
100 SHEETS
$435.00

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.23

$435.00


Available to ship on14 April 2025Details


Request more information

packaging

pack of 100 sheets

Quality Level

manufacturer/tradename

SureWick®

filter L × W

20 cm × 30 cm

thickness

0.83 mm

shipped in

ambient

General description

SureWick® Pad Materials are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. SureWick® cellulose fiber pad materials are used as sample and absorbant pads in lateral flow assays. This pack contains 100 sheets of 20cm x 30cm C083 cellulose fiber pad material.

Application

SureWick® Pad Materials are optimised for use as sample, conjugate, or absorbent pads in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.

Features and Benefits

  • SureWick® cellulose fiber pads are made from non-woven, 100% pure cellulose fiber.
  • These pads do not contain binders or glues, which can interfere with assay performance.
  • Manufactured to ensure excellent consistency, they provide reliable, predictable sample flow.
  • They exhibit consistent thickness, bed volume, and uniform surface quality.
  • Quick delivery and lead time.
  • Backed by excellent technical support.

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Velu Ranganathan et al.
Analytical biochemistry, 588, 113471-113471 (2019-10-16)
An aptamer-based colorimetric lateral flow assay was developed for the detection of human epidermal growth factor receptor 2 (HER2). In this study, two approaches were examined using HER2 binding aptamers and gold nanoparticles. The first method used was a solution-based
Lyubov V Barshevskaya et al.
Biosensors, 9(4) (2019-10-02)
An immunochromatographic test system has been developed for the simultaneous rapid multiplex serodiagnostics of bovine brucellosis, tuberculosis, and leukemia. The test system is based on the use of a conjugate of gold nanoparticles with the chimeric protein Cysteine-A/G and three
Aapo Knuutila et al.
Diagnostics (Basel, Switzerland), 10(4) (2020-04-02)
Most of the current serological diagnosis of pertussis is based on pertussis toxin (PT) IgG antibodies and does not differentiate between vaccination and infection-induced antibodies. PT is included in all of acellular pertussis vaccines available in the world. Multiplex testing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service