Skip to Content
Merck
All Photos(1)

Key Documents

CC305

Sigma-Aldrich

PureCol Coated 100-mm Dishes

Ultrapure collagen type I coated 100-mm dishes for cell culture applications (10 dishes)

Synonym(s):

Cell culture dishes, Petri dishes

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
$498.00

$498.00


Estimated to ship on25 May 2025

Available for sale now

Request a Bulk Order

Select a Size

Change View
10 EACH
$498.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$498.00


Estimated to ship on25 May 2025

Available for sale now

Request a Bulk Order

General description

Collagen is the main structural protein of the extracellular matrix found within various connective tissues of the body. Type I collagen is a major structural component of skin, bone, tendon, and other fibrous connective tissues. In order to provide the most in vivo-like environment for cells, researchers have historically cultured cells inside collagen hydrogels or on top of collagen coated plasticware.

PureCol® is an ultrapure bovine skin derived collagen that is approximately 97% Type I collagen with the remainder being comprised of Type III collagen. It contains a high monomer content as measured by gel permeation chromatography providing product consistency, reproducibility and is cited in over 2000 publications.

PureCol Coated T25 Flasks have a uniform and consistent application of high quality Type I collagen on clear polystyrene surface. The vented flasks have an anti-drip edge and modified canted neck to provide maximum control replacing nutrient depleted media. The flasks have an anti-tip feature and specially shaped pouring channel for improved handling. Flasks are aseptically processed and are non-pyrogenic.

Surface area/well: 58 cm2. Typical working volume: 10-20 mL. Coating concentration: 50-100 μg/mL.

PureCol is a registered trademark of Advanced BioMatrix, Inc.

Storage and Stability

Store PureCol Coated 10-mm dishes at room temperature.

Legal Information

PureCol is a trademark of Advanced BioMatrix, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service