Skip to Content
Merck
All Photos(1)

Documents

C5789-M

Sigma-Aldrich

Custom-OEM Cell Diss Soln (S-004), 100ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

Quality Level

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service