BX0865
Boric acid
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Synonym(s):
Boracic Acid
About This Item
Recommended Products
Agency
meets USP/NF
Quality Level
vapor pressure
2.6 mmHg ( 20 °C)
Assay
99.5% (alkalimetric)
form
crystals, granules or powder
technique(s)
electrophoresis: suitable
impurities
≤0.005% Insoluble in methanol
≤0.05% Nonvolatile with methanol
pH
3.8-4.8 (20 °C, 33 g/L in H2O)
mp
160 °C (dec.) (lit.)
solubility
water: 49.2 g/L at 20 °C
density
1.49 g/cm3 at 23 °C
anion traces
chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.01%
cation traces
Ca: ≤0.005%
Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%
shipped in
ambient
storage temp.
room temp
SMILES string
OB(O)O
InChI
1S/BH3O3/c2-1(3)4/h2-4H
InChI key
KGBXLFKZBHKPEV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
Calcium (Ca): 0.005% max
Chloride (Cl): 0.001% max
Form: Crystals or granular powder
Heavy metals (as Pb): 0.001% max
Insoluble in methanol: 0.005% max
Iron (Fe): 0.001% max
Nonvolatile with methanol: 0.05% max
Phosphate (PO4): 0.001% max
Sulfate (SO4): 0.01% max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service